Stable Isotopes
Scientist using stable isotope-labeled biomolecules in a lab.
Stable isotopes are non-radioactive forms of atoms. Their unique atomic structure and stability makes them useful as tracers in many compounds ranging from gases to complex molecules. Such labeled compounds are widely used in proteomics and metabolomics, agents for MRI / MRS, and in a broad range of other biomedical applications. 
We provide a sustainable, scalable, high-grade global supply of stable isotopes for research, medical, pharma, and electronics. In addition, we offer extensive product and packaging customization with fast turn-around, GMP capabilities, in-house technical expertise, and complete quality documentation.
Sort by Relevance
Showing 1-20 of 3967
Change View
Showing 1-20 of 3967
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 3967 results
Applications for Stable Isotopes
The use of our stable isotope-labeled compounds in metabolic and nutritional studies provides a means to dissect biochemical pathways. The availability of isotopically labeled compounds for basic scientific research permits the examination of the role of metabolic pathways in maintaining physiological states. In vivo studies using stable isotope labeled metabolites reveal important aspects of metabolism, including pathways and flux.
An important tool for high-resolution structure determination by NMR spectroscopy is the use of stable isotope-labeled biomolecules. By either selectively or uniformly incorporating stable isotopes into proteins, investigators can reduce the complexity of their spectra significantly. 13C, 15N, and deuterium are the most common isotopes incorporated into proteins through microbial expression systems, peptide synthesis, or with cell-free extracts.
The use of stable isotope compounds in medical imaging, in conjunction with 13C-MRS in vivo techniques, provides a method to explore fluxes through energy-related metabolic pathways for early diagnosis and treatment of neuropsychiatric disorders, cancer, and other diseases.
For these and many other applications, we offer a comprehensive portfolio of isotope labeled carbohydrates, amino acids, protected amino acids, metabolites, fatty acids, vitamins, steroids, other biologically-active compounds, and cell culture media and components. We also provide isotopically-labeled synthetic building blocks for researchers who prefer to make their own compounds. With our expertise in isotope synthesis, we stand ready to provide custom compounds for all of your specific needs.
Deuterated Products for Pharmaceuticals and Electronics
With nearly 4,000 isotopically labeled products, we provide unmatched reliability, flexibility, scalability, and support, making us the preferred partner for your needs at any stage of your project from research to production. 
Extensive Labeled Isotope Portfolio and Capacity
From milligram-quantities for drug discovery to metric-ton deliveries for manufacturing, our portfolio includes key products like Methanol-d4, Iodomethane-d3, and Benzene-d6, available in many grades and quantities to suit specific project requirements. Substantial capacity of D2, D2O, DMSO- d6, and many other products are available for projects of any size.
Robust Stable Isotope Supply Chain and Global Presence
We pride ourselves on our secure supply chain, which features capacity redundancy across multiple manufacturing sites and distribution centers around the world. This ensures that you have reliable access to materials, regardless of location. Our partnerships, including a supply contract with the India Heavy Water Board for D2O, further enhance our ability to deliver high-quality products consistently.
Isotope Customization and CMO Capabilities
We understand every project is unique. Our dedicated technical support team is here to assist you in customizing specifications and documentation to meet your exact needs from research materials to GMP. With strong global R&D teams with extensive process scale-up expertise, we can take your pharmaceutical intermediate or electronic precursor from bench to batch.
Expansion of infrastructure for key stable isotopes
We are committed to advancing the stable isotope availability for diagnostic, industrial and research applications. We recently built out our stable isotope center of excellence to increase production capacity by 50%, the GMP-grade urea-13C Active Pharmaceutical Ingredient (API) used by Urea-13C Breath Test (UBT) kit manufacturers to detect bacteria related to stomach disease.
Related Resources
Page 1 of 3