Figure 1. Cardiac troponin cTnC(1-89) in pLysS. Cells grown at 37 °C in shaker flasks. Red curve is ISOGRO supplemented minimal media and black curve is minimal media alone.
Figure 1. Cardiac troponin cTnC(1-89) in pLysS. Cells grown at 37 °C in shaker flasks. Red curve is ISOGRO supplemented minimal media and black curve is minimal media alone.
Figure 2. SDS-PAGE cTnC(1-89) cell lysates. M-low molecular weight marker; 0= uninduced; 1=induced minimal media; 2=induced minimal media + ISOGRO; L=Lysozyme. Data provided by Dr. Paul R. Rosevear The Department of Molecular Genetics, Biochemistry and Microbiology, University of Cincinnati Medical Center, Cincinnati, OH.
| 7 g/L Na2H PO4 | 1mg/L D-pantothenate |
| 3 g/L KH2PO4 | 1 mg/L biotin |
| 2.5 g/L NaCl | 50 mg/L thiamine |
| 10.5 g/L K2HPO4 | 1 mg/L pyridoxal phosphate |
| 0.5 g NaOH | 50 mg/L niacin |
| 1 g/L 15NH4Cl | 1 mg/L folic acid |
| 4 mM MgSO4 | 100 μg/L riboflavin |
| 10 mM FeCl3 | 1 mg/L choline chloride |
| 125 mM CaCl2 | 0.26 μg/L H3BO3 |
| 50 mM ZnSO4 | 2.4 ng/L Na2MoO4•2H2O |
| 2 g/L D-Glucose 13C6 | 16 ng/L CuCl2•2H2O |
| 107 μg/L MgCl2•6H2O | 0.16 μg/L MnCl2•4H2O |
| 20 μg/L FeCl2•4H2O |
Diminish lag time by > 60% and the number of hours required to attain high cell density by > 45% with the inclusion of 10% ISOGRO in M9.