Figure 1. The final yield of purified recombinant protein derived from each liter of culture. Acknowledgement: Data provided by Dr. Ross Overman and Dr. Kevin Embry, AstraZeneca, U.K.
Visit our ISOGRO Complex Growth Media page to find detailed information on products and protocols for using ISOGRO as a suppplement and as a stand alone media.
For optimal results as a stand-alone media, incorporate 10g of ISOGRO per Liter of culture.
Figure 1. The final yield of purified recombinant protein derived from each liter of culture. Acknowledgement: Data provided by Dr. Ross Overman and Dr. Kevin Embry, AstraZeneca, U.K.
A 39 μM sample of p38 alpha was produced from 50 mL of culture as seen below:
Figure 2. SDS-PAGE of p38 growth. Acknowledgement: Data provided by Dr. Jeffrey W. Peng, Dept. of Chem/Biochemistry, Univ. of Notre Dame, Notre Dame, Indiana
Figure 3. Cardiac troponin cTnC(1-89) in pLysS. Cells grown at 37 °C in shaker flasks. Red curve is ISOGRO supplemented minimal media and black curve is minimal media alone.
Figure 4. SDS-PAGE cTnC(1-89) cell lysates. Data provided by Dr. Paul R. Rosevear The Department of Molecular Genetics, Biochemistry and Microbiology, University of Cincinnati Medical Center, Cincinnati, OH.
Composition:
| Salts | 30% |
| Water | 3% |
| Glucose | 2% |
| Amino acids/Peptides | 65% |
Amino Acid Analysis:
| Ala | 13% | Ile | 4% | Pro | 4% | ||
| Arg | 3% | Leu | 8% | Ser | 4% | ||
| Asp | 14% | Lys | 6% | Thr | 5% | ||
| Glu | 10% | Met | 3% | Tyr | 3% | ||
| Gly | 12% | Phe | 4% | Val | 6% | ||
| His | 1% |