Stable Isotope Labeled Fatty Acids as Metabolic Tracers
Jennifer E. Lambert, Ph.D.
Parks Group
The University of Texas Southwestern Medical Center
Elizabeth J. Parks, Ph.D.
Associate Professor
Departments of Clinical Nutrition and Internal Medicine
The University of Texas Southwestern Medical Center
Postprandial Metabolism and Disease Risk
Traditionally, cardiovascular disease (CVD) risk factors have been determined and measured in the fasting state. However in recent years there has been an increasing awareness of the importance of elevated postprandial lipemia as an independent risk for CVD.1  Indeed, the efficiency with which the body manages incoming dietary macronutrients such as lipid can modulate disease risk in chronic conditions such as obesity2, type 2 diabetes,3,4 and non-alcoholic fatty liver disease.5  Interest in the postprandial state has led to the development of products and technologies designed to quantitate the kinetics of dietary fat (triacylglycerols, TG) absorption, assimilation, and disposal.  Development of isotope methodology has expanded the investigation of human lipid metabolism by allowing for a multitude of applications for measurement of in vivo lipid kinetics.  Although radioactive isotopes have been used to elucidate seminal findings in the area of fat metabolism,6 the present review will focus on the use of stablely-labeled compounds in human dietary research.
Applications of Stablely Labeled Dietary Tracers
The most obvious use of dietary lipid tracers is for investigation of lipid absorption after a test meal.  The labeled fat can be observed as lipids, fatty acids (FA) and TG, in plasma, chylomicrons (CM), and very low-density lipoprotein (VLDL) particles.  Dietary TG absorption reaches its maximum between 3-4 hours after the meal (Figure 1).  In studies of normal physiology, isotopically-labeled dietary FA can be used to determine if absorption depends on fat quality, such as structure of the FA7,8 or TG,9 and how absorbed FA are partitioned into lipid fractions other than TG.10,11  Alternatively, tracers can be used to determine if absorption is affected by diseases or stage of life.  Such applications have been used in studies of obesity,12,13 diabetes,4 malnourishment,14 and aging.15  In addition, because stable isotopes (and stable isotopically-labeled compounds) are naturally-occurring, these compounds are safe for use in many populations, including infants,16 children,14 pregnant or lactating women,7 and the elderly.15
The use of dietary tracers lends itself to a wide variety of analytical applications beyond the measurement of absorption.  Labeled precursors, such as the essential FA linoleic and linolenic acid, have allowed the determination of FA elongation and desaturation into longer-chain products.17,18  Combining tracer technologies with other compartment analyses can further expand the analytical possibilities of dietary tracers.  For example, tissue biopsies are used to assess tissue uptake of dietary fat into muscle, adipose, or liver.2,19,20  Lipid oxidation can be determined by measuring 13CO2 in breath or D2O in urine.8,21-23  Therefore, with the many applications possible, investigators can create and define new avenues of exploration in postprandial lipid metabolism.24
Fraction of Lipoprotein Chart
Figure 1
Healthy males (n=6, mean ± SEM) consumed two meals (0700 and 1200) of identical composition (32% fat, 54% carbohydrate, 14% protein), with each meal representing 1/3 of daily energy needs.  D31-tripalmitin (1.6g) was mixed with the liquid formula containing 40 g of dietary fat/meal as described previously.26, 30  Lipoprotein TG-palmitate was analyzed by GC/MS.  The d31-palmitate percentages (tracer/tracee) in the two lipoprotein fractions were compared to the TG enrichment in the liquid meal to calculate the percentage of lipoprotein-TG derived from the meal (Parks & Timlin unpublished data).
Practical Aspects and Study Design Considerations
Tracer Considerations
Common measurement techniques for analysis of metabolized labeled compounds are gas chromatography coupled with either mass spectrometry (MS) or isotope ratio mass spectrometry (IRMS).  Choice of analytical technique will depend on availability, cost, and sensitivity required for the experiment, and will influence the dose of tracer to be used.  For example, the higher sensitivity of IRMS allows for detection at lower levels of tracer enrichment; therefore smaller doses of tracer can be administered.
Tracer Form
As shown in Table 1, the choices of dietary tracers can include FA fed in the free form (e.g. Na+-palmitate) or as a TG (glyceryl trihexadecanoate-D31, or tripalmitin-D31).  The degree of isotope labeling will depend on the desired application, and cost, as some compounds are time-consuming to synthesize.  Tracers can be combined using different delivery methods in order to investigate multiple pathways of lipid metabolism.  Using the multiple stable isotope approach, Donnelly et al. utilized IV infusion of K+-13C4-palmitate on albumin to measure non-esterified FA metabolism, and a meal containing D31-tripalmitin to concurrently measure dietary FA metabolism to determine the lipid contributors to VLDL-TG.5  If multiple tracers are to be used, either different isotopic atoms should be chosen for each compound (e.g. Donnelly (2005) used a carbon-labeled FA and a deuterium-labeled TG) or it must be ensured that each tracer differs by enough mass to enable accurate and separate measurement of the products of interest.  For example, in order to investigate effects of sham feeding and multiple meals, Chavez-Jaurequi et al. incorporated 13C2-triolein into the evening meal and 13C7-triolein into the breakfast meal.13  Alternatively, different FA containing the same isotopic label (e.g. 13C-oleate and 13C-palmitate) can be used;25 however there is some debate as to whether this approach is appropriate, because differences in absorption8,11 or oxidation8,23 between different FA could influence results and interpretation.
Tracer Dose and Delivery
Dose of the tracer will depend on level of enrichment desired, metabolite of interest, and dietary fat content (Table 1A).  The tracer-to-tracee enrichment, or rather the comparison of amount of tracer FA fed to the amount of unlabeled dietary FA fed, can be as high as 10-20%.5,26  We have chosen a high level of enrichment in meals to be able to assess the early absorption of the label.  The ratio of the tracer (e.g. 100-800 mg) to the amount of total dietary fat (35-40 g) may be much lower (<3%) depending how much dietary fat is fed.25,27  The greater the amount fed in the meal, the more accurate assessment at earlier postprandial timepoints.
Capsules have been used previously to deliver tracer lipids,8,13 but it has been reported that the recovery of 13C in breath is 10 times higher when the tracer is fed in the meal compared to capsule form.23  This presents a problem for incorporation into a food if the tracer is a FA bound to a salt such as sodium or potassium; this is because the FA salt has physical properties as a soap.  Therefore, if the FA salt is solubilized in water, it must be handled gently or it will create foam and result in a food product more difficult to ingest as well as absorb.28  The incorporation of labeled FA into food (Table 1B) for administration has typically been achieved by emulsifying the FA into a sugar-protein complex, vegetable oil, or butter, and placed into food or a “milkshake” type beverage for consumption.10,11,29  The vehicle for the tracer often must be heated or sonicated in order to efficiently incorporate the tracer.  Barrows et al. used a homogenizer readily available in food processing labs (Model 110Y microfluidizer, Microfluidics, Newton, MA).30 Triglyceride tracers are more easily incorporated into test vehicles if the tracer is an oil (such as triolein), whereas tripalmitin is a crystal which must be heated for complete solubilization.
Methodological Considerations
Rate of Delivery and Meal Consumption
Early fat tolerance tests performed without isotopes were designed to provide a dose that would challenge the body’s lipid handling systems.31  Later, postprandial studies were designed to better mimic normal patterns of eating by including carbohydrate and protein with the fat,32,33 and feeding solid foods with the addition of isotopes to meals.  Investigators have used both liquid26 and solid5,22 meals (Table 1B).  Liquid meals have been provided as either continuous feedings via periodically sipping a beverage34 or by duodenal administration30 (Table 1B).  However, feeding the meal as a typical bolus is preferred because it produces greater label incorporation and greater fat oxidation8 compared to continuous feeding.30  The exception is studies in hospitalized infants, where set meal times are not practical.35 The choice of the amount of dietary fat fed in the test meal will depend on the increase in postprandial plasma lipids desired.  Increasing the amount of dietary fat leads to higher postprandial plasma TG concentrations,36 which dilutes the appearance of the label but can also amplify plasma lipid concentrations to a level sufficient for effective analysis.
Multiple Meals
Many investigators have used only one meal containing the tracer to measure postprandial metabolism. However, a delay in absorption has been demonstrated in the appearance of dietary tracer in the CM and VLDL TG pools13,25,26,37 (Figure 1).  The delay can occur from 4h to 13h after the first labeled meal.  This so-called “Second Meal Effect” is required to see maximum appearance of the dietary tracer in plasma lipid compartments,13,25 such that the consumption of a subsequent labeled meal is associated with more label appearance in blood (Figure 1).  After two meals, 100% of the CM-TG pool contains the same enrichment as the meal, leading to all of the TG in CM particles being derived from the tracer-containing meal.  This second meal does not need to contain fat, as a simple glucose drink has been shown to induce a CM lipid response.38  The Second Meal Effect also indicates that the food consumed the evening before the test influences postprandial metabolism.39 The pre-test meal composition should therefore be controlled by providing standardized meals at least the evening before, if not for 1-3 days leading up to the testing day.30  If using a 13C-labeled tracer, some investigators also instruct subjects to avoid foods naturally rich in 13C, such as corn products and cane sugar.4  Other lifestyle aspects to take into consideration include asking participants to avoid strenuous exercise and alcohol 2-3 days before the testing period.4,30
Table 1: Study design considerations
Key Study Design Consideration
A.    Tracer Aspects
Free fatty acid or Triglyceride
  • solubility in tracer vehicle
Other tracers used simultaneously
Dose
  • limit of analytical equipment
  • fat contained in the meal
Liquid vs. solid vs. capsule
B.    Dietary Aspects
Macronutrient composition of meal
  • amount of dietary fat
  • presence/absence of carbohydrate
Rate of delivery
  • continuous infusion
  • “sipping” (multiple small meals)
  • single meal bolus
Second meal effect
Future Directions
With the growing interest in postprandial metabolism and wide variety of applications, stable isotopes are an invaluable tool for investigating human metabolism.  A large body of literature utilizing isotopes to measure the absorption and disposal of dietary fat exists.  Standardized protocols have been published to aid researchers interested in studying the impact of dietary fat using stable isotopes.  Given the role of fat intake on the development of obesity and its related disorders, stable isotope and mass spectrometry methods can provide powerful tools for further characterization of normal and dysregulated physiological processes.  Such investigations will help to unravel how postprandial metabolism contributes to disease pathology and create novel clinical tools for screening and treatment.  Examination of postprandial lipid metabolism in obesity may pave the way for development of pharmaceutical or nutritional interventions for beneficial modification of postprandial lipemia to reduce disease risk.
Please visit the Metabolic Research webpage for a listing of related products.
Materials List
References
  1. Bansal S, Buring JE, Rifai N, Mora S, Sacks FM, Ridker PM. Fasting compared with nonfasting triglycerides and risk of cardiovascular events in women. JAMA. 2007; 298(3): 309-16.
  2. McQuaid SE, Hodson L, Neville MJ, Dennis AL, Cheeseman J, Humphreys SM, et al. Downregulation of adipose tissue fatty acid trafficking in obesity: a driver for ectopic fat deposition? Diabetes. 2011; 60(1): 47-55.
  3. Hodson L, Bickerton AST, McQuaid SE, Roberts R, Karpe F, Frayn KN, et al. The contribution of splanchic fat to VLDL triglyceride is greater in insulin-resistant than insulin-sensitive men and women: studies in the postprandial state. Diabetes. 2007; 56(10): 2433-41.
  4. Masding MG, Stears AJ, Burdge GC, Wootton SA, Sandeman DD. The benefits of oestrogens on postprandial lipid metabolism are lost in post-menopausal women with Type 2 diabetes. . Diabet Med. 2006; 23(7): 768-74.
  5. Donnelly KL, Smith CI, Schwarzenberg SJ, Jesserun J, Parks EJ. Sources of fatty acids stored in liver and secreted via lipoproteins in patients with nonalcoholic fatty liver disease. J Clin Invest. 2005; 115(5): 1343-51.
  6. Santosa S, Hensrud DD, Votruba SB, Jensen MD. The influence of sex and obesity phenotype on meal fatty acid metabolism before and after weight loss. Am J Clin Nutr. 2008; 88: 1134-41.
  7. Emken EA, Adlof RO, Hachey DL, Garza C, Thomas MR, Brown-Booth L. Incorporation of detuerium-labeled fatty acids into human milk, plasma, and lipoprotein phospholipids and cholesteryl esters. J Lipid Res. 1989; 30(3): 395-402.
  8. Jones PJH, Pencharz PB, Clandinin MT. Whole body oxidation of dietary fatty acids: implications for energy utilization. Am J Clin Nutr. 1985; 42(5): 769-77.
  9. Emken EA, Adlof RO, Duval SM, Shane JM, Walker PM, Becker C. Effect of triacylglycerol structure on absorption and metabolism of isotope-labeled palmitic and linoleic acid by humans. Lipids. 2004; 39(1): 1-9.
  10. Emken EA. Metabolism of dietary stearic acid relative to other fatty acids in human subjects. Amer J Clin Nutr. 1994; 60(6 Suppl): 1023S-8S.
  11. Hodson L, McQuaid SE, Karpe F, Frayn KN, Fielding BA. Differences in partitioning of meal fatty acids into blood lipid fractions: a comparison of linoleate, oleate and palmitate. Am J Physiol Endocrinol Metab. 2009; 296(1): E64-E71.
  12. Hodson L, McQuaid SE, Humphreys SM, Milne R, Fielding BA, Frayn KN, et al. Greater dietary fat oxidation in obese compared with lean men: an adaptive mechanism to prevent liver fat accumulation? Am J Physiol Endocrinol Metab. 2010; 299(4): E584-E92.
  13. Chavez-Jauregui RN, Mattes RD, Parks EJ. Dynamics of fat absorption and impact of sham feeding on postprandial lipema. Gastroenterology. 2010; 139(5): 1538-48.
  14. Murphy JL, Robinson EN, Forrester TE, Wootton SA, Jackson AA. Gastrointestinal handling and metabolic disposal of 13C-labelled tripalmitin during rehabilitation from childhood malnutrition. Br J Nutr. 2001; 85(6): 705-13.
  15. Puga GM, Meyer C, Everman S, Mandarino LJ, Katsanos CS. Postprandial lipemia in the elderly involves increased incorporation of ingested fat in plasma free fatty acids and small (Sf 20-400) triglyceride-rich lipoproteins. Am J Physiol Endocrinol Metab. 2011; 301(2): E356-E61.
  16. Lin YHL, A., Mena P, Uauy R, Salem N, Pawlosky RJ. Compartmental analyses of 2H5-alpha-linolenic acid and C-U-eicosapentanoic acid toward synthesis of plasma labeled 22:6n3 in newborn term infants. Amer J Clin Nutr. 2010; 92(2): 284-93.
  17. Emken EA, Adlof RO, Duval SM, Nelson GJ. Influence of dietary arachidonic acid on metabolism in vivo of 8-cis, 11-cis, 14-eicosatrienoic acid in humans. Lipids. 1997; 32(4): 441-8.
  18. Pawlosky RJ, Hibbeln JR, Lin Y, Goodson S, Riggs P, Sebring N, et al. Effects of beef- and fish-based diets on the kinetics of n-3 fatty acid metabolism in human subjects. Am J Clin Nutr. 2003; 77(3): 565-72.
  19. Bickerton AST, Roberts R, Fielding BA, Hodson L, Blaak EE, Wagenmakers AJM, et al. Preferential uptake of dietary fatty acids in adipose tissue and muscle in the postprandial period. Diabetes. 2007; 56(1): 168-76.
  20. Evans K, Burdge GC, Wootton SA, Clark ML, Frayn KN. Regulation of dietary fatty acid entrapment in subcutaneous adipose tissue and skeletal muscle. Diabetes. 2002; 51(9): 2684-90.
  21. Roberts R, Bickerton AST, Fielding BA, Blaak EE, Wagenmakers AJ, Chong MF, et al. Reduced oxidation of dietary fat after a short term high-carbohydrate diet. Am J Clin Nutr. 2008; 87(4): 824-31.
  22. Jones AE, Stolinski M, Smith RD, Murphy JL, Wootton SA. Effect of fatty acid chain length and saturation on the gastrointestinal handling and metabolic disposal of dietary fatty acids in women. Br J Nutr. 1999; 81(1): 37-43.
  23. DeLany JP, Windhauser MM, Champagne CM, Bray GA. Differential oxidation of individual dietary fatty acids in humans. Am J Clin Nutr. 2000; 72: 905-11.
  24. Parks E, Hellerstein MK. Carbohydrate-induced hypertriacylglycerolemia: An historical perspective and review of biological mechanisms. Amer J Clin Nutr. 2000; 71(2): 412-33.
  25. Heath RB, Karpe F, Milne RW, Burdge GC, Wootton SA, Frayn KN. Dietary fatty acids make a rapid and substantial contribution to VLDL-triacylglycerol in the fed state. Am J Physiol Endocrinol Metab. 2007; 292(3): E732-E9.
  26. Timlin MT, Barrows BR, Parks EJ. Increased dietary substrate delivery alters hepatic fatty acid recycling in healthy men. Diabetes. 2005; 54: 2694-701.
  27. Evans K, Burdge GC, Wootton SA, Collins JM, Clark ML, Tan GD, et al. Tissue-specific stable isotope measurements of postprandial lipid metabolism in familial combined hyperlipidemia. Atherosclerosis. 2008; 197(1): 164-70.
  28. Jones AE, Smith RD, Nurmi JA, Hameen-Anttila P, Murphy JL, Wootton SA. The gastrointestinal handling and postprandial metabolism of [1-13C]palmitic and [1-13C]oleic acids in healthy women. Proc Nutr Soc. 1997; 56: 197A.
  29. Murphy JL, Jones A, Brookes S, Wootton SA. The gastrointestinal handling and metabolism of 1-13C palmitic acid in healthy women. Lipids. 1995; 30(4): 291-8.
  30. Barrows BR, Timlin MT, Parks EJ. Spillover of dietary fatty acids and use of serum nonesterified fatty acids for the synthesis of VLDL-triacylglycerol under two different feeding regimens. Diabetes. 2005; 54: 2668-73.
  31. Ginsberg HN, Jones J, Blaner WS, Thomas A, Karmally W, Fields L, et al. Association of postprandial triglyceride and retinyl palmitate responses with newly diagnosed exercise-induced myocardial ischemia in middle-aged men and women. Arterio Throm Vasc Biol. 1995; 15: 1829-38.
  32. Redard C, Davis PA, Schneeman BO. Dietary fiber and gender, effects on postprandial lipemia. Am J Clin Nutr. 1990; 52: 837-45.
  33. Schneeman BO, Kotite L, Todd KM, Havel RJ. Relationships between the responses of triglyceride-rich lipoproteins in blood plasma containing apolipoproteins B-48 and B-100 to a fat containing meal in normolipidemic humans. Proc National Acad Sci. 1993; 90: 2069-73.
  34. Zheng C, Ikewaki K, Walsh BW, Sacks FM. Metabolism of apoB lipoproteins of intestinal and hepatic origin during constant feeding of small amounts of fat. J Lipid Res. 2006; 47(8): 1771-9.
  35. Sunehag AL, Haymond MW, Schanler RJ, Reeds PJ, Bier DM. Gluconeogenesis in very low birth weight infants receiving total parenteral nutrition. Diabetes. 1999; 48: 791-800.
  36. Dubois C, Beaumier G, Juhel C, Armand M, Portugal H, Pauli A-M, et al. Effects of graded amounts (0-50 g) of dietary fat on postprandial lipemia and lipoproteins in normolipidemic adults. Amer J Clin Nutr. 1998; 67: 31-8.
  37. Evans K, Kuusela PJ, Cruz ML, Wilhelmova I, Fielding BA, Frayn KN. Rapid chylomicron clearance following sequential meals:  effects of second meal composition. Brit J Nutr. 1998; 79: 425-9.
  38. Robertson MD, Parkes M, Warren BF, Ferguson DJP, Jackson KG, Jewell DP, et al. Mobilisation of enterocyte fat stores by oral glucose in humans. Gut. 2003; 52: 834-9.
  39. Robertson MD, Henderson RA, Vist GE, Rumsey RD. Extended effects of evening meal carbohydrate-to-fat ratio on fasting and postprandial substrate metabolism. Amer J Clin Nutr. 2002; 75: 505-10.
Related Links