Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
BisPhePhos XD gold(I) chloride: Efficient and Broadly Applicable Cationic Gold Catalyst
Introduction
Gold catalysis is a landmark addition to the field of organic synthesis,1–3 but the relatively large catalyst loading needed in many gold-catalyzed applications is impractical in large-scale synthesis. In order to make gold catalysis a better tool for synthetic chemists, the laboratories of GB Hammond and Bo Xu developed a broadly applicable gold precatalyst that can achieve high turnover (at ppm level gold loadings) under mild conditions (e.g. room temperature or slightly elevated temperature). Impressively, reactions were conducted in open-air containers using commercial solvents that needed no prior purification. Aldrich Chemistry is proud to offer this novel gold precatalyst, BisPhePhos XD gold(I) chloride (L511846),4 in collaboration with the Hammond (University of Louisville) and the Xu groups (Donghua University).
Advantages
  • Broadly applicable in a wide range of cationic gold-catalyzed reactions
  • Low catalyst loading (often at ppm level)
  • Mild reaction conditions (often room temperature)
  • Bench-top stable
Representative Schemes4
Special thanks to Professor GB Hammond and Professor Bo Xu for contributing this technology spotlight.