Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Rovis N-Heterocyclic Carbene (RoNHC) Catalysts
Sigma Aldrich is proud to offer the Rovis chiral N-heterocyclic carbene catalysts (RoNHCs) in partnership with Plura Biosciences.
The benefits of RoNHC technology:
  • Rapid synthesis of stereogenic fluorine centers with high ee
  • Metal-free, organocatalytic reactions
  • Simple room temperature procedures
α-Fluoroenals 1 (from 374717), when treated with the hydration (RoNHC-H) or amidation (RoNHC-A) catalyst afford enantioenriched α-fluorocarboxylic acids or enantioenriched α-fluoroamides in excellent yield and selectivity.
Available Building Blocks
Or, purchase your fluorine stereocenters pre-installed with our commercially available α-fluorocarboxylic acids.
Materials
To learn more about Professor Rovis’ research, please visit his Professor Product Page.
Related Links