The 3'/5' Assay for Analysis of RNA Integrity Protocol
Quantitative PCR Protocols
Although quantitative PCR uses the same basic concept as traditional PCR, the reactions differ in that the amplicons are generally smaller and are detected indirectly using an additional dye or labeled probe or primer.
The 3’/5’ integrity assay is a potential first step in the identification of RNA degradation. The assay is particularly useful when a large number of samples are to be analyzed or when the degradation is less than that detected by capillary systems but still sufficient to effect qPCR analyses.
One or more target RNA sequences are selected, and in this example GAPDH is used9. Two assays are designed along the length of the target such that one is located close to the 3’ UTR and the second is approximately 1 kb upstream. cDNA is generated using reverse transcription from an anchored oligo-dT primer. Following amplification by qPCR, the products from each assay are quantified and the ratio of the quantities is compared to that derived from a sample using high quality RNA. Degradation of the template results in a relative decrease in product, especially with assays near the 5’ end of the target gene. This results in an increase in the 3’/5’ ratio.
In the following test, the optimized conditions for the example assays are provided. It may be necessary to alter these conditions following optimization of assays to the targets that are more appropriate for the experiment (see Assay Optimization and Validation, Primer Concentration Optimization and Primer Optimization Using Temperature Gradient that describe the process of assay optimization and validation).
Equipment
  • Quantitative PCR instrument
  • Laminar flow hood for PCR set up (optional)
Reagents
Supplies
Table P9-23. Oligonucleotide Sequences for 3’/5’ Assay Using Human GAPDH as an Example.
Primer/Probe Sequence
GAPDH 5’ Forward GTGAACCATGAGAAGTATGACAAC
GAPDH 5’ Reverse CATGAGTCCTTCCACGATACC
GAPDH 3’ Forward AGTCCCTGCCACACTCAG
GAPDH 3’ Reverse TACTTTATTGATGGTACATGACAAGG
GAPDH 5’ Probe [FAM]CCTCAAGATCATCAGCAATGCCTCCTG[BHQ1]
GAPDH 3’ Probe [JOE] or [HEX] — instrument specific
CCCACCACACTGAATCTCCCCTCCT[BHQ1]
Method
1.    Place all the reaction components on ice to defrost.
2.    Mix and then centrifuge briefly to collect contents at the bottom of the tube.
3.    Calculate the number of reactions required, running two reactions per test sample and also two No Template Controls
       (NTC). Calculating the requirements for the number of reactions plus 10% extra to allow for pipetting error
       (Table P9-24).
4.    Prepare master mix that is sufficient for all samples and controls according to Table P9-24. Mix well and centrifuge
       briefly to collect contents at the bottom of the tube.
Table P9-24. qPCR Master Mix for the 3’/5’ Integrity Assay.
Reagents Volume (μL) per Single
25 μL Reaction
2× LuminoCt ReadyMix 12.5
Primer 3’ F (50 μM stock) 0.15
Primer 3’ R (50 μM stock) 0.15
Primer 5’ F (50 μM stock) 0.15
Primer 5’ R (50 μM stock) 0.15
Probe 3’ (10 μM stock) 0.5
Probe 5’ (10 μM stock) 0.5
Reference dye (optional) 1.0
PCR grade water 4.9
5.    Aliquot 20 μL of qPCR master mix into the PCR tubes/wells. If using a PCR plate, follow a plate schematic to
       ensure that the reagents and controls are added to the correct wells.
6.    Add 5 μL cDNA sample into qPCR tubes/wells.
7.    Cap tubes or seal the PCR plate and label. (Make sure the labeling does not obscure instrument
       excitation/detection light path.)
8.    Samples should be run according to the three-step protocol provided in Table P9-25. Following the initial hot
       start, steps 1–3 are repeated through 40 cycles.
Note: The conditions on Table P9-25 are optimal for the human GAPDH example assay given. However it may be appropriate to run alternative assays using the two-step protocol as described in Table P9-25B.
Table P9-25. qPCR Cycling Conditions for the 3’/5’ Integrity Assay.
Cycling Conditions Temp (°C) Time (sec)
Initial Hot Start/denaturation 95 30
Steps 1–3 are repeated through 40 cycles
Step 1 95 5
Step 2
55 15
Step 3
72 10
Table P9-25B. Fast PCR Cycling Conditions.
FAST Cycling Conditions Temp (°C) Time (sec)
Initial Hot Start/denaturation 95 30
Steps 1 and 2 are repeated through 40 cycles
Step 1 95 5
Step 2
60 30
Materials