Enzymatic Assay of Carboxypeptidase A
1. OBJECTIVE
To standardize a procedure for the assay of Carboxypeptidase A at Sigma-Aldrich, St. Louis.
2. SCOPE
This procedure applies to all products that have a specification for Carboxypeptidase A including, but not limited to, Carboxypeptidase A from Bovine Pancreas, Sigma-Aldrich Product Number C9268.
3.1 Purified Water = water from a deionizing system, resistivity > or = 18MΩ•cm @ 25ºC
3.2 Carboxypeptidase A Unit Definition – One unit will hydrolyze 1.0 μmole of hippuryl-L-phenylalanine per min at pH 7.5 at 25ºC.
4. DISCUSSION
Hippuryl - L- Phenylalanine Carboxypeptidase A > Hippuric acid + Phenylalanine
5. RESPONSIBILITIES
It is the responsibility of trained Analytical Services laboratory personnel to follow this procedure as written.
6. SAFETY
Refer to Material Safety Data Sheets (MSDS) for hazards and appropriate handling precautions.
7.1 CONDITIONS:
T = 25°C, pH = 7.5, A254nm, Light path = 1 cm
7.2 METHOD:
Continuous Spectrophotometric Rate Determination
7.3 REAGENTS:
7.3.1 25 mM Tris HCl Buffer with 500 mM Sodium Chloride, pH 7.5 at 25°C (Buffer)
Prepare a solution in purified water containing 3.03mg/ml Trizma Base, such as Sigma-Aldrich Product Number T1503, and 29.2mg/ml Sodium Chloride, such as Sigma-Aldrich Product Number S9888. Adjust the pH to 7.50 at 25°C using 1N Hydrochloric Acid. Keep solution at room temperature.
7.3.2 1.0 mM Hippuryl-L-Phenylalanine Solution (Hippuryl-L-Phe)
Prepare a solution in 200 Proof Ethanol containing 32.6 mg/ml Hippuryl-L-phenylalanine, such as Sigma-Aldrich Product Number H6875. Once the sample is completely in solution in ethanol, further dilute the solution in Reagent 7.3.1 (Buffer) to 0.326 mg/ml. Do not adjust the pH of this solution. Keep the solution at room temperature and use within three hours of preparation.
7.3.3 1.0 M Sodium Chloride Solution (Enzyme Diluent)
Prepare a solution in purified water containing 58.4 mg/ml Sodium Chloride, such as Sigma-Aldrich Product Number S9888. Keep solution at room temperature.
7.3.4 Carboxypeptidase A Enzyme Solution (Enzyme)
7.3.4.1 Before use, prepare a solution in room temperature Reagent 7.3.3 (Enzyme Diluent) containing 4-8 units/ml of Carboxypeptidase A.
7.3.4.2 Do not dilute the enzyme in cold Reagent 7.3.3.
7.3.4.3 Make a fresh dilution of enzyme in reagent 7.3.3 (Enzyme Diluent) for each aliquot.
7.3.4.4 Perform independent dilutions for each sample and repeat to verify consistency of aliquots taken from the retainer or bulk.
7.3.4.5 Prior to pulling a sample, be sure to shake the sample to insure all material that has settled to the bottom is in a homogeneous solution.
7.4 PROCEDURE
7.4.1 Run one aliquot at a time because the maximum rate is typically in the first minute.
7.4.2 Blank the spectrophotometer against reagent 7.3.2. Because of the high absorbency of reagent 7.3.2, the assay needs to be run on instrument that is accurate above an absorbency of 2.0.
7.4.3 Pipette the following (in milliliters) into suitable quartz cuvettes:
| Test | Blank | |
| Hippuryl-L-Phe (Reagent 7.3.2) | 2.90 | 2.90 |
7.4.4 Equilibrate to 25°C using a suitably thermostatted spectrophotometer. Then add (in milliliters):
| Enzyme Diluent (Reagent 7.3.3) | ---- | 0.10 |
| Enzyme (Reagent 7.3.4) | 0.10 | ---- |
7.4.5 Mix by inversion and record the increase in absorbance at 254nm for approximately 5 minutes. Obtain the fastest linear rate (ΔA254nm/minute) over a one minute interval for the test and the blank reactions. The ΔA254nm/minute must fall between 0.05 and 0.1 for each reaction for data to be valid.
7.5 CALCULATIONS
| 7.5.1 | Units/ml enzyme = | (ΔA254nm/min Test - ΔA254nm/min Blank)(3.00)(df) |
| (0.36)(0.10) |
| 7.5.2 | Units/mg solid = | Units/ml enzyme |
| mg solid/ml enzyme |
| 7.5.3 | Units/mg protein = | Units/ml enzyme |
| mg protein/ml enzyme |
3.00 = Total volume (in milliliters) of assay
df = Dilution Factor
0.36 = Millimolar Extinction Coefficient of hippuric acid at 254nm
0.10 = Volume (in milliliters) of enzyme used
7.6 FINAL ASSAY CONCENTRATION :
In a 3.00 ml reaction mix, the final concentrations are 24 mM Tris, 0.96% ethanol, 0.97 mM hippuryl-L-phenylalanine, 512 mM sodium chloride, and 0.4-0.8 units Carboxypeptidase A.
8.1 Bergmeyer, H.U., Gawehn, K., and Grassl, M. (1974) Methods of Enzymatic Analysis (Bergmeyer, H.U., ed.) Volume 1, 2nd ed., 436-437, Academic Press, Inc. New York, NY
8.2 Replaces Enzyme OP SPHPHE01 Version 1
Review, approvals and signatures for this document will be generated electronically using EDMS. Print a “For Use” copy if hardcopy with signature verification is required.