Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Enzymatic Assay of Elastase (EC 3.4.21.36)
Description
This procedure may be used for Elastase products using SucAla3-pNA as the substrate.
The continuous spectrophotometric rate determination (A410, Light path = 1 cm) is based on the following reaction:
where:
SucAla3-pNA = N-Succinyl-Ala-Ala-Ala-p-nitroanilide
SucAla3 = N-Succinyl-Ala-Ala-Ala
p
NA = p-Nitroanilide
Unit Definition – One unit of Elastase will hydrolyze 1.0 µmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per minute at pH 8.0 at 25 °C.
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Reagents and Equipment Required
Trizma® base (Catalog No. T1503)
N-Succinyl-Ala-Ala-Ala-p-nitroanilide (Catalog No. S4760)
Preparation Instructions
Use ultrapure water (≥18 MΩxcm resistivity at 25 °C) for the preparation of reagents.
Buffer (100 mM Tris HCl, pH 8.0 at 25 °C) – Prepare a 12.1 mg/mL solution of Trizma base (Catalog No. T1503) in ultrapure water. Adjust the pH to 8.0 at 25 °C with 1 M HCl.
Substrate Solution (4.4 mM SucAla3-pNA Solution) – Prepare 2 mg/mL solution of N-Succinyl-Ala-Ala-Ala-p-nitroanilide (Catalog No. S4760) in Buffer.
Enzyme Solution (Elastase) – Immediately before use, prepare a solution containing 0.2–0.5 unit/ml of Elastase in cold (2–8 °C) buffer.
Procedure
In a 3.00 ml reaction mix, the final concentrations are 96.7 mM Trizma, 0.29 mM N-Succinyl-Ala-Ala-Ala-p-nitroanilide, and 0.02–0.05 unit of Elastase.
1.     Pipette the following reagents into suitable cuvettes:
Reagent Test (ml) Blank (ml)
Buffer 2.70 2.80
Substrate Solution 0.20 0.20
2.     Mix by inversion and equilibrate to 25 °C. Then add:
Reagent Test (ml) Blank (ml)
Enzyme Solution 0.10
3. Immediately mix by inversion and record the increase in A410 for ~5 minutes. Obtain the ΔA410/minute using the maximum linear rate for both the Test and Blank using a minimum of 4 data points over a one minute time interval.
Results
Calculations
1.
Units/ml enzyme = (ΔA410/min Test – ΔA410/min Blank) (3.00) (df)
(8.8) (0.10)
where:
3.00 = Total volume (ml) of assay
df = Dilution factor
8.8 = Millimolar extinction coefficient of p-Nitroaniline at 410 nm at pH 8.0
0.1 = Volume (ml) of Enzyme Solution used
2.
Units/mg solid = units/ml enzyme
mg solid/ml enzyme
Materials
References
  • Bieth, J. et al., Biochemical Medicine, 11, 350-357 (1974).
Trizma is a registered trademark of Sigma-Aldrich Co. LLC.
02/14-1
Related Links