Enzymatic Assay of Mutanolysin
Description
This procedure may be used for Mutanolysin products. The Continuous Spectrophotometric Rate Determination (A600, Light path = 1 cm) is based on the following reaction:
Unit Definition: One unit of Mutanolysin will produce a ΔA600 of 0.01 per minute at pH 6.0 at 37 °C in a 1 ml volume using a suspension of Streptococcus faecalis cell walls as the substrate.
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Reagents and Equipment Required
MES hydrate (Catalog No. M8250)
1.0 M Magnesium chloride solution (Catalog No. M1028)
TES (Catalog No. T1375)
Mutanolysin Assay Substrate (Catalog No. M3440)
Preparation Instructions
Use ultrapure water (≥18 MΩxcm resistivity at 25 °C) for the preparation of reagents.
Buffer (50 mM MES Buffer with 1 mM magnesium chloride, pH 6.0, at 37 °C) – Dissolve 1.95 grams of MES hydrate (Catalog No. M8250) in 190 ml of ultrapure water and then add 0.2 ml of 1.0 M Magnesium chloride solution (Catalog No. M1028). Adjust the pH to 6.0 at 37 °C using 1 M NaOH and then bring the volume of the entire solution to 200 ml using ultrapure water.
Enzyme Diluent (50 mM TES with 1 mM magnesium chloride, pH 7.0 at 37 °C) – Dissolve 2.29 grams of TES (Catalog No. T1375) in 190 ml of ultrapure water and then add 0.2 ml of 1.0 M Magnesium chloride solution (Catalog No. M1028). Adjust the pH to 7.0 at 37 °C using 1 M NaOH and then bring the volume of the entire solution to 200 ml using ultrapure water.
Substrate (Streptococcus faecalis Cell Wall Suspension) – Prepare a cell wall suspension of log-phase Streptococcus Faecalis STF-3 (ATCC® 12784) cells in ~10 ml of Buffer to a concentration where the A600 is in the range of 0.48–0.52. Alternatively, Mutanolysin Assay Substrate (Catalog No. M3440) may be prepared according to the instructions in the product datasheet.
Enzyme Solution (Mutanolysin) – Immediately before use, prepare a solution containing 500–700 units/ml of mutanolysin in cold (2–8 °C) Enzyme Diluent.
Procedure
In a 3.04 ml reaction mix, the final concentrations are 49 mM MES, 1 mM MgCl2, 0.66 mM TES, and 6.5–9.2 units of mutanolysin.
1. Pipette the Substrate into suitable cuvettes.
Reagent Test
(ml)
Blank
(ml)
Substrate 3.00 3.00
2. Equilibrate at 37 °C and monitor the A600 until constant using a suitably thermostatted spectrophotometer. Then add:
Reagent Test
(ml)
Blank
(ml)
Enzyme Solution
0.04
Enzyme Diluent 0.04
3. Immediately mix by inversion and record the decrease in A600 for 10 minutes. Using a 1 minute time period and a minimum of 4 data points, obtain the ΔA600/minute using the maximum linear rate for both the Test and the Blank.
Results
Calculations
1.
Units/ml enzyme = (ΔA600/min Test – ΔA600/min Blank) (3.04) (df)
(0.01) (0.04)
where:
3.04 = Volume (ml) of reaction mix
df = Dilution Factor
0.01 = Decrease in absorbance per minute as defined by the unit definition
0.04 = Volume (ml) of Enzyme Solution used
2.
Units/mg solid = units/ml enzyme
mg solid/ml enzyme
Materials
 Reference
  • Calandra, G.B., and Cole, R.M., Infection and Immunity, 28, 1033-1037 (1980).
ATCC is a registered trademark of American Type Culture Collection.
03/14-1
Related Links