Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Escort™ IV Transfection Reagent Protocol
Product No. L3287
Introduction
Escort IV is a liposome suspension composed of a polycationic lipid and a neutral, non-transfecting lipid compound.  Transfection using liposomes is a commonly used method for the introduction of DNA into eukaryotic cells. This technique has been used to obtain both transient and stable transfections in a wide variety of cell types (see below). The procedure is based on the formation of a complex between the plasmid DNA and the lipid reagent, which adheres to the cell surface and is taken up by the cell, presumably by endocytosis, releasing the DNA into the cytoplasm.
Cells tested for good efficiency transfection
Mammalian cell lines Insect Cells Primary cells
C2C12
CHOK1
COS-1
COS-7
HEK293
HeLa
LNCaP
NIH3T3
T-24
S2 (D. mel.)
S2R+
Sf-9 (adherent)
Sf-9 (suspension)
T. Ni.
Aortic smooth muscle (rat)
Bone marrow endothelial cells (human)
Keratinocytes (human)
Myoblasts (rabbit)
Storage
2 – 8°C.  DO NOT FREEZE.
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Procedure for transfecting adherent primary cells and cell lines
The following protocol is designed for good efficiency across a variety of cell types. For highest transfection efficiency, use the Optimization Protocol to determine the best set of conditions for your cells.
Day One: Plate Cells
Plate the cells one day prior to transfection. Use a seeding density that will provide >70% confluence about 12 hours later, generally according the chart below:
Culture Dish Total sells to plate (per well)
24 well plate 0.5 – 2 x105
12 well plate 0.25 - 1 x106
6 well plate or 3.5 cm dish 0.5 – 2 x106
10 cm dish 1 - 2 x106
Day Two: Transfection
1. Dilute DNA and lipid reagent for complexing according to the chart below. Use only unsupplemented basal medium for dilution. In tube A, dilute the plasmid DNA in medium. In tube B, dilute the lipid reagent in medium.
  Tube A Tube B  
Culture Dish DNA volume (µL) Medium volume (µL) Escort IV Transfection Reagent volume (µL) Medium volume (µL) Approximate volume of media during transfection
24 well plate
0.5
9.5
0.5
9.5
500 µL
12 well plate 1
49
1
49
1 mL
6 well plate or 3.5 cm dish 2
98
2
98
2 mL
10 cm dish 5
495
5
495
5 mL
2. Add tube A to tube B and mix gently. Allow complexes to form 15-30 minutes at room temperature.
3. Remove half the medium from each cell culture dish and discard.
4. Add complexes drop-wise to the cell cultures.  Swirl the plates gently to distribute the complexes evenly over the cells.
5. Incubate the cells 5-18 hours at 37°C, and then change the medium (use regular growth medium). Allow the cells in continue incubating 24 – 72 more hours.
Day Three and beyond: Analyze Cells
Collect and lyse the cells – they are ready to be used for other applications. Alternatively, you can passage the cells for use in other applications.