Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Initiating a Starter Culture
  1. Pick a single colony from a freshly streaked plate and inoculate a starter culture with 3 to 5 mL of media.
  2. Use the appropriate antibiotic and incubate at 37 °C* for approximately 8 hours while shaking at 250-300 rpm.
  3. Use the starter culture to inoculate an overnight culture. Dilute the starter culture 1:500 to 1:1000 in a large flask with the appropriate volume of media and incubate at 37 °C* for 12 to 16 hours while shaking at 250-300 rpm.
*This temperature is for standard E. coli. Some bacteria require different culture temperatures.
Materials
Categories