Protein Determination Modified Lowry Method
1. OBJECTIVE
To standardize a procedure for the determination of protein by modified Lowry at Sigma-Aldrich, St. Louis, MO.
2. SCOPE
This procedure applies to all products that have a specification for Lowry protein determination without Trichloroacetic Acid (TCA) precipitation.
Purified water - from a deionizing system, resistivity > or = 18 MΩ•cm @ 25°C
| Copper + Protein | Alkaline pH > | Copper-protein complex |
| Phenol reagent |
5. RESPONSIBILITIES
It is the responsibility of all Analytical Services laboratory personnel to follow this procedure as written.
6. SAFETY
Refer to the Material Safety Data Sheet (MSDS) for hazards and appropriate handling precautions.
7.1 CONDITIONS:
T = 25°C, A750nm, Light path = 1 cm
7.2 METHOD:
Colorimetric
7.3 REAGENTS:
7.3.1 0.85% Sodium Chloride Solution (NaCl) Use Sodium Chloride Solution, 0.85%, Sigma-Aldrich Product Number S0817 , or prepare 100 ml in purified water using Sodium Chloride, Sigma-Aldrich Product Number S9888 .
7.3.2 0.01% Lowry Protein Standard (WPS) Prepare 10 ml in Reagent 7.3.1 using Sigma-Aldrich Product Number P0914 or a working protein standard prepared per Sigma “Preparation of Working Protein Standard for Use in Protein Determination” (Operating Procedure 10-30-6802).
7.3.3 Modified Lowry Reagent (LOW) Prepare by reconstitution in purified water, per label instructions, a vial of Lowry Reagent, Modified, Sigma Product Number L1013 orSigma Product Number L3540 . This solution can be stored for approximately one month or until the solution becomes cloudy.
7.3.4 Protein Sample Solution (Sample) Prepare a solution containing 10-100 mg protein/ml of sample in reagent 7.3.1.
7.3.5 330 M Folin and Ciocalteu’s Phenol Reagent (F&C) Prepare by dilution an appropriate volume of Folin and Ciocalteu’s Phenol Reagent, Sigma-Aldrich Product Number F9252 in purified water and mix thoroughly. Prepare fresh immediately prior to use.
7.4 TEST METHOD
Inhibition Reaction
Pipette (in milliliters) the following reagents into a suitable containers:
| Test | Std 1 | Std 2 | Std 3 | Std 4 | Std 5 | Blank | |
| Reagent 7.3.1 (NaCl) | ----- | 0.90 | 0.70 | 0.50 | 0.30 | ----- | 1.00 |
| Reagent 7.3.2 (WPS) | ----- | 0.10 | 0.30 | 0.50 | 0.70 | 1.00 | ----- |
| Reagent 7.3.3 (LOW) | 1.00 | 1.00 | 1.00 | 1.00 | 1.00 | 1.00 | 1.00 |
| Reagent 7.3.4 (Sample) | 1.00 | ----- | ----- | ----- | ----- | ----- | ----- |
Mix thoroughly by vortexing and incubate for 10 minutes at 25C. Incubation time must be at least 10 minutes to allow for the copper solution to react with peptide bonds. This time can be as long as 24 hours if needed.
Then add:
| Test | Std 1 | Std 2 | Std 3 | Std 4 | Std 5 | Blank | |
| Reagent 7.3.5 (F&C) | 0.50 | 0.50 | 0.50 | 0.50 | 0.50 | 0.50 | 0.50 |
Mix thoroughly by vortexing and incubate for 30 minutes at 25°C. Incubation time must be at least 30 minutes but no longer than 60 minutes, because color development will continue after 30 minutes. All standards, samples, and blank absorbances must be read on spectrophotometer within a short time of each other.
Transfer the solutions to suitable cuvettes and record the A750nm for Test, Standards, and Blank using a suitable spectrophotometer.
7.5 CALCULATIONS
ΔA750nm Standard = A750nm Standard – A750nm Blank
Prepare a standard curve by plotting the ΔA750nm of the Standards vs. mg of protein.
Sample Determination:
ΔA750nm Test = A750nm Test - A750nm Blank
Determine the mg of protein from the Standard Curve.
mg Protein = (mg of protein from the Standard curve) * (df)
df = Dilution factor
| % Protein = | (mg Protein) * (100) |
| (mg solid/ml) |
100 = Conversion to percentage
| mg Protein/ml | (mg Protein) |
| (ml sample/ml) |
8.1 Lowry, O.H., Rosebrough, N.J., Farr, A.L and Randall, R.J. (1951) J. Biol. Chem. 193, 265-275
8.2 Peterson, G.L. (1979) Analytical Biochemistry, 100, 201-220
| Print Name | Sign Name | Title | Date | |
| Prepared by: | Holly Chapman | Holly Chapman | Originator | 11/9/04 |
| Approved by: | Rodney Burbach | Rodney Burbach | Supervisor, Analytical Services | 11/10/04 |
| Approved by: | Gene McNaughton | Gene McNaughton | Quality Assurance | 11/10/04 |