Anti-Digoxigenin-AP, Fab fragments Protocol & Troubleshooting
Product No. 11093274910
Protocol
Substrates for AP
  • CSPD and CDP- Star
  • NBT/BCIP
  • Fast Red Tablets
  • BM-Purple
Troubleshooting
DIG Application Manual for Nonradioactive In Situ Hybridization 4th Edition, page 32, Correction for 50× Denhardt’s
50× Denhardt’s:
1% Ficoll (type 400), 1% polyvinylpyrrolidone, and 1% bovine serum albumin.