Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Biotin-16-dUTP Protocol & Troubleshooting
Product No. 11093070910
Protocol
PCR-labeling with Biotin-16-dUTP
The incorporation of Biotin-16-dUTP using PCR is possible with most DNA polymerases, e.g., as described for DIG-dUTP in the Package Insert of Taq DNA Polymerase.
More information is available in the Roche PCR Manual and the Roche DIG Application Manual for Filter Hybridization.
Troubleshooting
Evaluation of labeling efficiency
There is no biotin-labeled control DNA available. For a rough estimation of labeling efficiency, use a dilution series in a direct spot test, similar to the method used to evaluate DIG-labeled probes.
After estimating the labeling concentration set up a dilution series of the labeled DNA probe. For blocking of the membrane, use 5% Blocking Reagent.
Instead of the anti-DIG antibody Streptavidin-AP (alkaline phosphatase) conjugate, Cat. No. 11 093 266 910, use a 1:5000 dilution for direct colorimetric or luminescent detection of the biotin-labeled DNA probe spotted on the membrane.
Materials
Liberase is a trademark of Roche.
cOmplete is a trademark of Roche.
Related Links