Blocking Reagent Protocol & Troubleshooting
Product No. 11096176001
Protocol
Preparation of stock solution (10x):
Blocking Reagent is dissolved in Maleic acid buffer [100 mM Maleic acid, 250 mM NaCl, pH 7.5 (20 °C) adjust with conc. or solid NaOH], to a final concentration of 10% (w/v) with shaking and heating either on a heating block to +60 °C for approx. 1 h, or until the reagent is completely in solution, or in a microwave oven 3 to 4 min total. The Blocking Reagent must be heated while dissolving, but must not be boiled. Boiling causes the reagent to coagulate. When the Blocking Reagent does not go into solution, check pH of solution, and adjust if necessary, and continue heating.
Note: Dilute the 10x Blocking Solution with Maleic acid buffer to a 1x Blocking Solution. Always prepare freshly!
Blocking Reagent and Whole Mounts
During the in situ hybridization procedure, nonspecific background is more efficiently reduced by adding Roche tRNA or Roche fish sperm DNA to the hybridization solution.
The Blocking Reagent is used to decrease the background in nonradioactive hybridization and detection of nucleic acid hybrids: 1) Blocking prior to the detection procedure is optional; either use the blocking reagent, or the appropriate serum from the animal (e.g., sheep) from which the antibody was obtained. Antibodies may be pre-adsorbed for 1 hour using a whole mount specimen to reduce the background.
Protocol for "Use in Immunological Detection in ISH"
For details, refer to DIG-RNA probes & tissue sections in the Roche Non-radioactive In situ Hybridization Manual.
After last washes, continue to immunological detection:
Place slides in a tray suitable for 8 slides with 30 ml of buffer.
Incubate as follows (and change trays rather than just solutions and wash the extra set of trays between
incubations to avoid background):
a) Buffer 1 [100mM Tris-HCl pH7.5, 20 °C; 150 mM NaCl] 5 minutes
b) Buffer 2 [buffer 1 with 0.5% (w/v) Blocking Reagent] 1 h
c) Buffer 3 [buffer 1 with 1% (w/v) BSA; 0.3% (v/v) Triton X-100] 1 hour
d) Drain excess buffer 3 (do not let the tissue become dry), and add 100 μl buffer 4 [buffer 3 with anti-DIG-AP 1:1000 to 1:3000 depending on your target) on the tissues
PREPARE FRESHLY BEFORE USE.
e) Incubate 1 hour at RT in a moist chamber
f) Wash 4 times in buffer 3 (20 minutes)
g) Equilibrate in buffer 1 for 5 minutes
h) Incubate 5 minutes in buffer 5 (100 mM Tris-HCl (pH 9.5, 20 °C; 100 mM NaCl; 50 mM MgCl2);
i) Perform the color reaction (e.g., NBT/BCIP) in the dark; cover trays to avoid evaporation; check in appropriate time intervals; stop enzyme reaction and wash off background.
Microarrays
In general, Roche recommends using arrays with an epoxy or any other non-charged surface to keep background signals as low as possible.
Most important is appropriate blocking prior to addition of the fluorophore-conjugated anti-DIG. Roche recommends using the blocking buffer included in the DIG Wash and Block Buffer Set.
First, wash the array after hybridization, and then preblock using a solution of 1% Blocking Reagent in 2x PBS containing 0.1% Tween 20. Dilute the dye-labeled anti-DIG in 1% Blocking Reagent in 2x PBS containing 0.1% Tween 20 and apply it to the array. For washing use 2x PBS, 0,1% Tween 20. Alternatively, a buffer containing 1% BSA instead of the DIG Bblocking Rreagent or 5% Denhardt 's reagent can be used.
Troubleshooting
Increased concentration, up to 5%
The concentration of blocking reagent can be increased up to 5%. This can have a positive effect in reducing background staining. For most standard DIG applications, the recommended concentration is sufficient. For biotin detection, use a concentration of 5%.
The easiest way to use the Blocking Reagent is with the Roche DIG Wash and Block Buffer Set (Cat.No. 01585762001), a ready-to-use set of stock solutions.
High Background
The reason is possibly nonspecific sticking of the labeled DNA to proteins or other DNA-binding molecules in the tissue. To reduce high background staining, follow advice given in the protocol help corner, in particular consider pre-adsorbing the antibody mix.