Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glutamate Dehydrogenase (GlDH) Protocol
Product No. 10197734001
Protocol
Formulation of Glutamate Dehydrogenase
Glutamate Dehydrogenase is a component of cofactor recycling systems for NAD(P) and NAD(P)H. The formulation contains buffer substances, BSA as stabilizer and ADP as activator; please note that ADP can produce an adsorption peak at 260 nm. Less than <0.005% contaminants, such as ADH, LDH, and MDH each, may be present. For additional additives and contaminants, please refer to the Certificate of Analysis. Less than <0.005% contaminants, such as ADH, LDH, and MDH each, may be present.
For best results, dissolve the lyophilizate in redistilled water.
Stability of Glutamate Dehydrogenase
  1. Lyophilized material: Stable at +2 to +4 °C, when stored dry. A decrease in activity of approx. 10% can occur within one month, when stored dry at room temperature (+15 to +25 °C).
  2. Solution: Dissolving contents of the vial (3,000 U) in 2 ml redistilled water results in a solution with a pH of approx. 8. GIDH is very unstable at pH <5 or >8.
A 10% decrease in activity can occur within 7 days when stored at +2 to +8 °C, or within one day when stored at room temperature (+15 to +25 °C). There is no significant decrease in activity, when this solution is frozen or lyophilized.