Glycogen Protocol & Troubleshooting
Product No. 10901393001
Protocol
Recovery of RNA:
For quantitative recovery of low concentrations of RNA (ng/ml), an inert coprecipitant (e.g., glycogen, yeast RNA, or linear acrylamide) should be used.
Troubleshooting
Diluted samples:
At < 0.02 mg/ml DNA/RNA recovery may be low. To improve recovery of diluted samples, add carrier (glycogen, tRNA or sperm DNA) to sample.