Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Neuraminidase (Sialidase) Protocol
Product No. 10269611001
Protocol
Using Neuraminidase to remove sialic acid from purified lipooligosaccharide (LOS)
The enzyme can be used to cleave sialic acids off proteins:
A mixture of N-glycosidase F, O-glycosidase, and neuraminidase is often useful (O-glycans often have sialidized structures which could be hydrolyzed by neuraminidase and then O-glycosidase could continue hydrolyzing the structure).
The enzyme/substrate ratio should be approx. 0.04 U/25 - 80 μg.
Combination with O-Glucosidase:
To use neuraminidase from Vibrio cholerae (instead of neuraminidase from Arthrobacter) together with O-glycosidase under the same conditions, one should take into account the pH-optima of O-glycosidase and the Vibrio neuraminidase, i.e., one should work at pH 6 instead of pH 7.2.
Treatment of formalin-fixed cells with neuraminidase
The following procedure adapted from Corral et al (1990) BBRC 172:1349-1356:
  • Wash 3x107 cells in Dulbecco′s PBS
  • Fix cells for 1 h at +4 °C in an equal amount of 1% paraformaldehyde, 0.1M sodium-cacodylate, pH 7.3
  • Wash cells three times with PBS and resuspend them in PBS at a concentration of 3x107 cells /ml
  • Use 150μl of this cell suspension and treat for 1h at +37 °C with 250U/ml neuraminidase from Vibrio cholerea
Specific inhibitors of neuramindase (silaidase) from Vibrio cholerae:
  • 2,3-Dehydro-2-deoxy-N-acetylneuraminic acid
  • Higher homologues of the dehydro-deoxy-N-acyl-neuraminic acid series (acyl = propionyl...)
  • Di-isopropyl-fluoro-phosphate
It has not been determined whether SDS, guanidine HCl, or urea inhibit the enzyme, but inactivation is possible.
It has not been determined whether CHAPS, Triton X100/X114, ß-mercaptoethanol, or Manidet P40 inhibit the enzyme, but most likely no inhibition occurs. No information is available on inhibition by DTT.