Subculture of Adherent Cell Lines
Cook Book Sept 2010 Volume 12, Fundamental Techniques in Cell Culture Laboratory Handbook-2nd Edition
12.4 Protocol 3 - Subculture of Adherent Cell Lines
back to top
Aim
Adherent cell lines will grow in vitro until they have covered the surface area available or the medium is depleted of nutrients. At this point the cell lines should be sub cultured in order to prevent the culture dying. To subculture the cells they need to be brought into suspension. The degree of adhesion varies from cell line to cell line but in the majority of cases proteases, e.g. trypsin, are used to release the cells from the fl ask. However, this may not be appropriate for some lines where exposure to proteases is harmful or where the enzymes used remove membrane markers/receptors of interest. In these cases cells should be brought into suspension into a small volume of medium mechanically with the aid of cell scrapers.
back to top
Materials
  • Media– pre-warmed to 37°C (refer to the ECACC Cell Line Data Sheet for the correct medium)
  • 70% (v/v) isopropanol in sterile water
  • PBS without Ca2+/Mg2+
  • 0.25% trypsin/EDTA in HBSS, without Ca2+/Mg2+
  • Soyabean Trypsin Inhibitor
  • Trypan blue (vital stain)
back to top
Equipment
  • Personal protective equipment (sterile gloves, laboratory coat, safety visor)
  • Waterbath set to appropriate temperature
  • Microbiological safety cabinet at appropriate containment level
  • Incubator
  • Pre-labelled flasks
  • Inverted phase contrast microscope
  • Centrifuge
  • Haemocytometer
  • Marker Pen
  • Pipettes
  • Ampoule rack
  • Tissue
back to top
Procedure
  1. View cultures using an inverted microscope to assess the degree of confl uency and confi rm the absence of bacterial and fungal contaminants.
  2. Remove spent medium.
  3. Wash the cell monolayer with PBS without Ca2+/Mg2+ using a volume equivalent to half the volume of culture medium. Repeat this wash step if the cells are known to adhere strongly.
  4. Pipette trypsin/EDTA onto the washed cell monolayer using 1ml per 25cm2 of surface area. Rotate flask to cover the monolayer with trypsin. Decant the excess trypsin.
  5. Return fl ask to the incubator and leave for 2-10 minutes.
  6. Examine the cells using an inverted microscope to ensure that all the cells are detached and floating. The side of the flasks may be gently tapped to release any remaining attached cells.
  7. Resuspend the cells in a small volume of fresh serum-containing medium to inactivate the trypsin. Remove 100-200μl and perform a cell count (see Protocol 6 - Cell Quantifi cation). In the case of cells cultured in serum-free media, use a trypsin inhibitor e.g. soyabean trypsin inhibitor to inactivate the trypsin.
  8. Transfer the required number of cells to a new labelled fl ask containing pre-warmed medium (refer to ECACC Cell Line Data Sheet for the required seeding density).
  9. Incubate as appropriate for the cell line.
  10. Repeat this process as demanded by the growth characteristics of the cell line.
back to top
Key Points
  1. Some cultures whilst growing as attached lines adhere only lightly to the fl ask, thus it is important to ensure that the culture medium is retained and the fl asks are handled with care to prevent the cells detaching prematurely.
  2. Although most cells will detach in the presence of trypsin alone the EDTA is added to enhance the activity of the enzyme.
  3. Trypsin is inactivated in the presence of serum. Therefore, it is essential to remove all traces of serum from the culture medium by washing the monolayer of cells with PBS without Ca2+/Mg2+.
  4. Cells should only be exposed to trypsin/EDTA long enough to detach cells. Prolonged exposure could damage cell surface receptors.
  5. Trypsin should be neutralised with serum prior to seeding cells into new flasks otherwise cells will not attach.
  6. Trypsin may also be neutralised by the addition of soyabean trypsin inhibitor, where an equal volume of inhibitor at a concentration of 1mg/ml is added to the trypsinised cells. The cells are then centrifuged, resuspended in fresh culture medium and counted as above. This is especially necessary for serum-free cell cultures.
  7. If a CO2 incubator is not available gas the fl asks for 1-2min with 5% CO2 in 95% air filtered through a 0.2μm filter.
  8. If the cells harvested are at too low a cell density to re-seed at the appropriate cell density into fresh flasks it may be necessary to centrifuge the cells e.g. 5 mins at 150 x g, and resuspend in a smaller volume of medium.
back to top
Did You Know?
Serum-free media will not inactivate trypsin. A trypsin inhibitor, e.g. soyabean trypsin inhibitor, must be used.