Testing for Mycoplasma by Culture Isolation
Cook Book Volume 12
Fundamental Techniques in Cell Culture Laboratory Handbook-2nd Edition
Flow Scheme for Detection of Mycoplasma by Culture
Aim
Detection of mycoplasma by culture is the reference method of detection and has a theoretical level of detection of 1 colony-forming unit (cfu). However, there are some strains of mycoplasma that are non-cultivable (certain strains of Mycoplasma hyorhinis). The method is suitable for the detection of mycoplasma in both cell cultures and cell culture reagents and results are obtained within 4 weeks. Mycoplasma colonies observed on agar plates have a ‘fried egg’ appearance (see figure 1).
Figure 1 Typical “fried egg colonies” Mycoplasma pneumoniae (magnification x400)
Materials
- 70% (v/v) isopropanol in sterile water
- Mycoplasma plates (in 5cm petri dishes)
- Mycoplasma horse serum broths (in 1.8ml aliquots)
- M. orale (NCTC* 10112)
- M. pneumoniae (NCTC* 10119)
*Available from the National Collection of Type Cultures (NCTC)
Equipment
- Personal protective equipment (sterile gloves, laboratory coat, safety visor)
- Waterbath set to 37°C
- Microbiological safety cabinet at appropriate containment level
- Incubator set at 37°C
- Anaerobic jar system
Procedure
- Inoculate 2 agar plates with 0.1ml of test sample.
- Inoculate an agar plate with 100 cfu of each control organism.
- Leave 1 agar plate un-inoculated as a negative control.
- Inoculate 1 broth with 0.2 ml of test sample.
- Inoculate a broth with 100 cfu of each control organism.
- Leave 1 broth un-inoculated as a negative control.
- Incubate agar plates anaerobically for 14 days at 37°C.
- Incubate broths aerobically for 14 days at 37°C.
- Between days 3 - 7 and 10 - 14 of incubation, subculture 0.1 ml of test broth onto an agar plate and incubate plate anaerobically as above.
- Observe agar plates after 14 days incubation at x400 magnifi cation using an inverted microscope for the presence of mycoplasma colonies (see Figure 1).
Criteria for a Valid Result
All positive control agar plates and broths show evidence of mycoplasma by typical colony formation on agar plates and usually a colour change in broths. All negative control agar plates and broths show no evidence of mycoplasma.
Criteria for a Positive Result
Test agar plates infected with mycoplasma show typical colony formation.
Criteria for a Negative Result
The test agar plates show no evidence of mycoplasma.
Notes
- Mycoplasma colonies have a typical colony formation commonly described as “fried egg” due to the opaque granular central zone of growth penetrating the agar surrounded by a flat translucent peripheral zone on the surface.
- Mycoplasma pneumoniae is a potential pathogen and must be handled in a class 2 microbiological safety cabinet operating to ACDP Category 2 Conditions.
- This test procedure should be carried out in a microbiology laboratory away from the cell culture laboratory.
- ECACC recommends that samples are tested for mycoplasma using at least two detection methods (e.g. indirect DNA stain and culture isolation) for a more reliable result. This is due to the varying detection sensitivities of the methods for different species of mycoplasma.
- ECACC also offers a mycoplasma screening PCR assay which can return a result within 24 hours.