Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Stille Coupling in a Box
Product No. 802336
Reagents
Compound Catalog No. Equivalents MW Density Amount mmol
Kit Components
Pd[P(t-Bu)3]2 676578 0.03 511.05   7.7 mg 0.015
DABCO D27802 3.0 112.17   170 mg 1.50
NaCl S9888 1.0 58.44   30 mg 0.50
TPGS-750-M 733857     1.002 2.0 mL  
User Supplied
Aryl bromide [ArBr]   1.0       0.50
Organotributyltin reagent [RSnBu3]   1.1       0.55
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Procedure
  1. Add the aryl bromide (0.5 mmol) and the organotributyltin reagent (0.55 mmol) into the provided 5-mL reaction vial containing a stir bar and preloaded catalyst Pd[P(t-Bu)3]2 (7.7 mg, 0.015 mmol), DABCO (170 mg, 1.50 mmol) and NaCl (30 mg, 0.50 mmol) under N2.
  2. Replace the solid top cap with the provided septum top cap and connect with N2 bubbler through a needle.
  3. Transfer the provided TPGS-750-M/H2O (2.0 mL; 2 wt %) solution into the reaction vial using the provided syringe.
  4. Turn on the stirrer and stir the resulting mixture vigorously at room temperature until the reaction is complete (monitored by TLC, GC, LC, etc.).
  5. After completion, dilute the reaction mixture with 1-3 mL of EtOAc (or i-PrOAc) followed by gently stir for 2 minutes and then filter the entire mixture through a bed of silica gel plug (1 inch) on a sintered glass Buchner funnel (30 mm diameter).
  6. Wash the silica gel bed with an additional 1 mL of EtOAc (or i-PrOAc) and collect all of the coupled material.
  7. Remove all of the volatiles in vacuo and purify the crude material by flash column chromatography on silica gel.
More information on the research from the Lipshutz Group can be found in the Professor Product Portal.  
Additional information on TPGS-750-M: Second Generation Amphiphile for Organometallic Chemistry in Water at Room Temperature can be found in the Technology Spotlight section of our website.