Core functionalization of Polyester-32-hydroxyl-1-acetylene bis-MPA dendron
Product Description
Theoretical MW: 3655.64 g/mol
Average Molecular Formula: C158H252O94
Number of hydroxyl groups: 32
Number of acetylene groups: 1
This monodisperse compound has multiple surface hydroxyl groups and a single alkyne focal point function. The alkyne function is well suited for “Click chemistry” catalytic cycloaddition to azides (see Procedure). The hydroxyl groups are selectively reactive with anhydrides or acyl halides. It is transparent to light in the UV/Vis wavelength region.
The product is a freeze-dried, white powder, which may contain traces of methanol or ether.
back to top 
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
The product is soluble in polar organic solvents (THF, DMSO, DMF, and methanol) and water.
back to top 
Storage/Stability
Polyester dendrons are susceptible to hydrolysis, which is accelerated when material is dissolved in water, at acidic/basic pH, or at elevated temperatures. For extended storage, store the dry powder at 2–8 °C.
back to top 
Procedure
Core functionalization via “Click chemistry”
(see Figure 1)
  1. Dissolve the dendron in THF or DMF and add a minimum of 1.2 equivalents of the appropriate azido derivative (see available azides at www.sigma-aldrich.com/click) per equivalent of acetylene functionality.
  2. Add water until the solution becomes opaque.
  3. Add 1 equivalent of (+)-sodium L-ascorbate (Catalog Number A7631) and 0.5 equivalent of copper(II) sulfate (Catalog Number 451657).
  4. Follow the reaction with 1H NMR until the peak corresponding to the acetylene proton disappears (a triplet at ca 2.43 ppm; C≡CH, 1H).
  5. When the reaction is complete, evaporate all the solvents and purify the compound by gradient flash chromatography (EtOAc       > EtOAc:MeOH) to obtain the final product.
Figure 1. Reaction Scheme (686611)
back to top 
Related Products
Polyester-8-hydroxyl-1-acetylene bis-MPA dendron, generation 3 (Catalog Number 686646 )
Polyester-16-hydroxyl-1-acetylene bis-MPA dendron, generation 4 (Catalog Number 686638 )
Manufactured by Polymer Factory Sweden AB.
back to top 
Materials
References
  1. Malkoch, M., et al., Multivalent, Bifunctional Dendrimers Prepared by Click chemistry. Chem. Communications, 46, 5775-5777 (2005).