Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
EDC-mediated couplings of Polyester-16-hydroxyl-1-carboxyl bis-MPA dendron
Product Description
Theoretical MW: 1,759.7 g/mol
Average Molecular Formula: C75H125O46
Number of hydroxyl groups: 16
Number of carboxylic acid groups: 1
This monodisperse compound has multiple surface hydroxyl groups and a single carboxylic acid focal point function. The carboxylic acid function is well suited for EDC-mediated coupling to amine-containing reagents. The hydroxyl groups are selectively reactive with anhydrides or acyl halides (see Figure 1). It is transparent to light in the UV/Vis wavelength region.
Figure 1. Reaction Scheme (686662)
The product is a freeze-dried, white powder, which may contain traces of methanol or ether.
back to top 
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
The product is soluble in polar organic solvents (THF, DMSO, DMF, and methanol) and water.
back to top 
Storage/Stability
Polyester dendrons are susceptible to hydrolysis, which is accelerated when material is dissolve d in water, at acidic/basic pH, or at elevated temperatures. For extended storage, store the dry powder at 2–8 °C.
back to top 
Related Products
Polyester-8-hydroxyl-1-carboxyl bis-MPA dendron, generation 3 (Catalog Number 686670)
Polyester-32-hydroxyl-1-carboxyl bis-MPA dendron, generation 5 (Catalog Number 686654)
EDC, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride (Catalog Number E1769)
Manufactured by Polymer Factory Sweden AB.
back to top 
Materials
References
EDC-mediated couplings:
  1. Desai, M.C., and Stramiello, L.M.S., Polymer Bound EDC (P-EDC): A Convenient Reagent for Formation of an Amide Bond. Tetrahedron Letters, 34, 7685-7866 (1993).
  2. Xu, Y., and Miller, M.J., Total Syntheses of Myobactin Analogues as Potent Antimycobacterial Agents Using a Minimal Protecting Group Strategy. J. Org. Chem., 63, 4314-4322 (1998).
  3. Futami, J. et al., Preparation of Potent Cytotoxic Ribonuecleases by Cationization. Enhanced Cellular Uptake and Decreased Interaction with Ribonuclease Inhibitor by Chemcial Modification of Carboxyl Group. Biochemistry, 40, 7518-7524 (2001).