Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Negative Photoresist Procedure
Introduction
The Aldrich photoresist kit is designed to have the necessary chemical components for each step in the lithographic process. The component materials are provided in pre-weighed quantities for your convenience. Etchants are available separately so that the proper etchant can be chosen for a variety of substrate choices.
back to top 
Handling and Storage of photoresist materials:
Precautions should be taken so no agglomeration forms within the photoresist materials:
  1. The kit should be refrigerated to inhibit polymerization and warmed to room temperature before opening.
  2. Kit materials should be stored away from light sources and kept in the sealed bottles.
back to top 
Procedure conditions:
Any resist procedure should be performed only under the proper conditions:
  1. Caution should be used to avoid dust or other particles.
  2. The relative humidity should be controlled between 30-50%.
  3. Appropriate lighting, preferrably of gold fluorescent, yellow incandescent, or white fluorescent with yellow or orange filters should be used.
Adequate ventilation is necessary when using any of the photoresist kit components.
back to top 
Outline of Photoresist Procedure:
  1. Substrate Preparation Clean and dry 20-30 min. at 120-130°C
  2. Photoresist Application Best with a spin-coating system
  3. Prebake 20 min. at 82°C
  4. Photoresist Exposure 1-10 sec. minimum light source 10mW/cm2
  5. Photoresist Develop 10-60 sec. spray (651788 Negative resist developer I) followed by isopropyl alcohol rinses and drying with compressed air or nitrogen.
  6. Postbake Minimum 10 min. 120°C (max. 148°C)
back to top 
Detailed Procedure
  1. Substrate Preparation:
    It is very important that the substrate is free of all organic residues and surface particles removed. Cleaning is accomplished by a thorough solvent rinse followed by baking at temperatures between 120-200°C for up to 20 minutes. Recommended solvent is trichloroethylene.
  2. Application:
    A spin-coating system is recommended for achieving a highly uniform and reproducible coating thickness. The substrate should be coated with the photoresist prior to spinning. Never apply the resist while spinning as an uneven distribution may result. Square or rectangular substrates are best coated at low rpm (50-1000 rpm). Undiluted resist at 75 rpm yields about 2.5 μm coatings, the substrate edges and corners being thicker. Resist thickness is dependent on the spin rate and acceleration and the viscosity of the resist. Circular substrates are best coated at a high rpm (2,000-5,000 rpm). Spinning of the substrate at >5,000 rpm has little effect on the thickness of the resist. As viscosity decreases the dependence of thickness on spin rate also decreases. In order to achieve a more uniform and thinner film, and to avoid thickening at the substrate edge, acceleration to peak rpm of 0.1 sec is optimum. Film thickness varies from 0.3 to 2 μm using a spin-coat system. Thick films (1-2 μm) are generally preferred in order to avoid etch penetration and the formation of pinholes. Thick films, however, can result in a loss of resolution. For better results, two thin coats can be applied.
  3. Prebake:
    In order to achieve the maximum adhesion of the resist to the substrate, all the residual solvent and volatile components should be removed by evaporation. Failure to thoroughly bake the resist can inhibit the crosslinking necessary for the resist process. Caution should be taken to avoid excessive baking that can result in fogging or decomposition of the resist. Recommended bake-time is 20 min @ 82 °C Baking above 104 °C will adversely affect adhesion.
  4. Photoresist Exposure:
    Any light source with near-UV emissions can be used to expose the photoresist. Large-area light sources should be used only for coarse lines (50 μm or larger). A less diffused light source is necessary for fine line patterns. Recommended fine-line pattern sources are carbon, high-pressure mercury vapor or xenon flash lamps. Proper exposure requires light intensities of ca. 100 mW/cm2. Thickness and processing variable can affect exposure. Exposure times of 1-10 seconds are typically sufficient, providing the light source yields a minimum irradiation of 10 mW/cm2 at the substrate surface. Exposure energy should not vary by more than 10% of optimum value or fine-line definition and reproducibility can be lost. Diffraction effects of the photoresist can cause cross-linking under the mask resulting in line broadening by as much as 2.5 μm.
  5. Photoresist Development:
    Spray the developer on to the coated substrate for 10-60 seconds followed by a rinsing several times with isopropyl alcohol. The surface should then be dried by blowing nitrogen or pure, compressed air over it.
  6. Postbake:
    A postbake of the photoresist is required to remove residual solvents and volatile components and helps enhance the chemical stability and adhesion of the resist. The recommended postbake is 10-20 min. at 120 °C and should never exceed 148 °C.
  7. Photoresist Removal:
    Hot chlorinated hydrocarbons, such as those in 651761 Negative resist remover I, swell the photoresist which, in conjunction with hot H2SO4 effectively remove the resist film. Ideal temperature for the remover and H2SO4 is 50-60 °C.
    back to top 
Materials
Product #
Image
Description
Molecular Formula
Add to Cart
654892   Aldrich® Negative Photoresist Kit I  
651826   Chromium etchant standard  
651478 Gallium antimonide (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm GaSb
651486 Gallium arsenide (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm AsGa
651494 Gallium phosphide (single crystal substrate), <111>, diam. × thickness 2 in. × 0.5 mm GaP
651842   Gold etchant, nickel compatible  
651818   Gold etchant, standard  
651796 Negative photoresist I  
651788   Negative resist developer I  
651761   Negative resist remover I  
651974   Negative resist thinner I  
651834   Nichrome etchant standard  
646687 Silicon wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm Si
647535 Silicon wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm Si
647780 Silicon wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm Si
647802 Silicon wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm Si
647675 Silicon wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm Si
647764 Silicon wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm Si
647101 Silicon wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm Si
647543 Silicon wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm Si
647799 Silicon wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm Si
647705 Silicon wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm Si
647772 Silicon wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm Si
Related Links