Preparation of Bottom-Gate OFET using P3HT
Product Description
Plexcore OS 1100, a p-type polymer semiconductor, is a moderate molecular weight grade of P3HT that is optimized for Organic Field Effect Transistor (OFET) applications. It has been used successfully in top and bottom-gate OFET architectures with a variety of device dimensions, substrates, and dielectrics. For bottomgate OFET, SiO2 is typically encountered in the open literature when a silicon wafer is used as the gate electrode. A number of other dielectrics are available for use with other substrates. Top-gate OFET devices require rather unique organic dielectrics that do not affect the organic semiconductor during its processing.
Plexcore OS 1100 has metal impurities less than 50 ppm. The specific impact of metallic impurities is dependent on which metals are present and at what concentration. There isn’t a significant impact on OFET performance due to metal impurities at this stated level, although this does not rule this possibility out due to end-use application.
Purity : 99.995% (trace metals basis)
head-to-tail regioregular : >95% (1H-NMR)
Fluorescence properties : lex 450 nm; lem 575 nm
Typically, higher molecular weight results in higher hole carrier mobilities for OFETs. The cause of this effect is not well understood.
Average Mn : 25,000–35,000 Mw/Mn <2
Mobility is extracted from the OFET transfer curve in the saturation region (i.e., from the slope of ÖIds vs. Vg curve) and is impacted by device architecture. In general, mobilities are higher for smaller OFET channel lengths. However, different device architectures will have behavior unique to that particular design.
back to top Back to Top
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
Typically, aromatic and halogenated aromatic solvents are used. The product is soluble in trichlorobenzene, chlorobenzene, toluene, and xylenes. In addition, chloroform and, to a lesser degree, tetrahydrofuran (THF), are both options.
back to top Back to Top
Storage/Stability
Store the product at room temperature.
In general, conjugated organic semiconductors (like Plexcore OS) may be sensitive to some (higher energy) wavelengths of light in the presence of oxygen. While this is not an acute problem for the bulk material, solutions and thin films tend to show some sensitivity over time. Therefore, storage and handling in an inert atmosphere (like nitrogen or argon) is recommended. A yellow filter light is a standard feature for processing these materials outside of a glove box. A combination of inert atmosphere and yellow light filter is best.
back to top Back to Top
Procedure
Note: A yellow filter light is a standard feature for processing these materials outside of a glove box. Storage and handling in an inert atmosphere (like nitrogen or argon) is recommended.
Typical Bottom-Gate OFET Architecture (see Figure 1):
Substrate : Silicon/SiO2 wafer
Gate : Silicon
Dielectric : SiO2 treated with HDMS
Organic Semiconductor : Plexcore OS 1100
Source/Drain : Au/Ti
Figure 1. Schematic of OFET using Plexcore OS 1100
Poly(3-hexylthiophene-2,5-diyl) (Plexcore OS 1100) solution:
Prepare a 5–20 mg/ml solution in an appropriate solvent or solvent blend (chloroform, toluene, xylene, chlorobenzene, o-dichlorobenzene, trichlorobenzene). Shake the solution at 40 °C (until well dissolved). Filter the solution with a 0.2 mm PTFE syringe filter.
Substrate Preparation:
Prior to spin coating, typical substrate preparation includes sonication with solutions such as distilled water, detergent water, isopropyl alcohol (electronic grade). After sonication, the substrate should be dried with a nitrogen gun.
Film Deposition via Spin Coating:
Using a filter syringe, apply the organic semiconductor onto the substrate surface. Using a spin coater, spread the solution at 350 rpm for 5 seconds and spin at 600–2,000 rpm for ~2 minutes. This will require optimization based on spin coating equipment and device architecture.
Drying Conditions:
Anneal at 100–175 °C for 5–60 minutes. Plexcore is a registered trademark of Plextronics, Inc. Product of Plextronics, Inc. U.S. Patent 6,166,172.
back to top Back to Top