Sigma-Aldrich®
Product Portfolio
|
|
|
Sigma-Aldrich® gives you easy access to the product you need
- by streamlining our product range
- by delivering clearly defined product categories
- by providing brands and grades for every application
Sigma-Aldrich Brands
PRODUCT BRANDS |
 |
| |
| APPLICATIONS |
Analytical Capillary Electrophoresis Chromatography Electrochemistry Microbiology Spectroscopy Titration / HYDRANAL® Wet Analysis |
Life Science Biochromatography Biotransformation Cell Culture Cloning & Expression Electrophoresis Enzymatic Analysis Fluorescence Spectroscopy Light & Electron Microscopy Mass Spectrometry Nucleic Acid Purification Protein Purification & Detection
|
Chemistry Asymmetric Synthesis Catalysis Inorganic Chemistry Medicinal Chemistry Organic Synthesis Organometallic Chemistry
Materials Science Alternative Energy Biomaterials Metal & Ceramic Science Micro/NanoElectronics Nanomaterials Organic Electronics Polymer Science |
Development and Manufacturing Biosciences Diagnostics High Technology Flavors & Fragrances Pharma Raw Materials for Manufacturing |
Sigma-Aldrich Grades
| Application |
Brand |
Grade or Trade Name |
Definition |
| Analytical |
|
TraceSELECT® Ultra |
Highest analytical grade for ultra-trace analysis. Designed for the ultra-trace analysis at ppb and even ppt levels. Typical metal trace impurities are below 0.1 µg/kg (<0.1 ppb), in some cases even as low as the ppq level. |
|
| Analytical |
|
TraceSELECT® |
High purity reagents for inorganic trace analysis applications and organic residue analysis. Developed for sample preparation and analysis in the ppm and ppb level. The blank values for metal traces are typically below 0.01 mg/kg (0.01 ppm). |
|
| Analytical |
|
TraceCERT® |
Certified reference material according to ISO 17025 and ISO Guide 34. TraceCERT reference materials are developed and produced in an accredited laboratory fulfilling both ISO 17025 (general requirements for the competence of testing and calibration laboratories) and ISO Guide 34 (general requirements for the competence of reference material producers). |
|
| Life Science |
|
BioUltra |
Finest quality products for life science, with the most stringent testing protocols and highest purity. Superior manufacturing methods and product specific testing protocols are designed to ensure quality to our highest standard. Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing. |
|
| Life Science |
|
BioXtra |
Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo. Includes application-specific formulations, such as tablets and prepared solutions. |
|
| Life Science |
|
BioReagent |
Base grade products, suitability tested for specific applications such as molecular biology or cell culture. |
|
| Life Science |
|
% purity |
Sigma brand products used for basic life science applications. |
|
| Chemistry |
|
Anhydrous |
High-purity solvents with extremely low water content specifically manufactured for moisture sensitive syntheses. |
|
| Chemistry |
|
% purity |
High quality chemistry products defined by their assay value as a percent purity. |
|
| All |
|
CHROMASOLV® |
Solvents tested for suitability in HPLC applications. Highest quality solvents used in LC-MS and LC-NMR are branded Fluka. |
|
| All |
 |
puriss p.a., ACS reagent |
Exceeds standard ACS specifications with additional trace metals analysis. |
|
| All |
 |
ACS reagent |
Meets or exceeds ACS specifications. For general analysis and procedures requiring stringent quality specifications. |
|
| All |
 |
Corresponds to official monograph (e.g. Ph.Eur., USP) |
Produced/procured, packaged and analyzed under appropriate quality systems for pharmaceutical applications according to the corresponding monograph. |
|
| All |
 |
Meets analytical specifications of USP, FCC, NF, EP (Ph. Eur.), BP, and/or JP. |
Chemicals that meet the latest testing specifications of the United States Pharmacopeia (USP), Food Chemicals Codex (FCC), National Formulary (NF), Pharmacopeia Europe (EP or Ph. Eur.), British Pharmacopeia (BP), and/or Japanese Pharmacopeia (JP). For R&D and testing purposes only. |
|
| All |
 |
ReagentPlus® or Laboratory reagent |
≥95% for general laboratory use. |
|
| All |
 |
Technical |
Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%. Variation in color and physical form is possible. |
©2010 Sigma-Aldrich Co. All rights reserved. SIGMA, SAFC, SIGMA-ALDRICH, ALDRICH, FLUKA, and SUPELCO are trademarks belonging to Sigma-Aldrich Co. and its affiliate Sigma-Aldrich Biotechnology, L.P. Sigma brand products are sold through Sigma-Aldrich, Inc. Sigma-Aldrich, Inc. warrants that its products conform to the information contained in this and other Sigma-Aldrich publications. Purchaser must determine the suitability of the product(s) for their particular use. Additional terms and conditions may apply. Please see reverse side of the invoice or packing slip. TraceSELECT, TraceCERT, CHROMASOLV, Hydranal and ReagentPlus are registered trademarks of Sigma-Aldrich Biotechnology L.P. and Sigma-Aldrich Co. BCR and ERM are registered trademarks of the European Commission.