Sigma-Aldrich
Technical Service > Product Portfolio
Sigma-Aldrich®
Product Portfolio
Sigma-Aldrich® gives you easy access to the product you need
  • by streamlining our product range
  • by delivering clearly defined product categories
  • by providing brands and grades for every application
Sigma-Aldrich Brands
PRODUCT
BRANDS
Fluka® Logo
Supelco® Logo
Sigma® Life Science Logo Aldrich® Logo
Aldrich Materials Science Logo
SAFC® Logo
APPLICATIONS Analytical
Capillary Electrophoresis
Chromatography
Electrochemistry
Microbiology
Spectroscopy
Titration / HYDRANAL®
Wet Analysis
Life Science
Biochromatography
Biotransformation
Cell Culture
Cloning & Expression
Electrophoresis
Enzymatic Analysis
Fluorescence Spectroscopy
Light & Electron Microscopy
Mass Spectrometry
Nucleic Acid Purification Protein Purification & Detection
Chemistry
Asymmetric Synthesis
Catalysis
Inorganic Chemistry
Medicinal Chemistry
Organic Synthesis
Organometallic Chemistry
Materials Science
Alternative Energy
Biomaterials
Metal & Ceramic Science
Micro/NanoElectronics
Nanomaterials
Organic Electronics
Polymer Science
Development and Manufacturing
Biosciences
Diagnostics
High Technology
Flavors & Fragrances
Pharma
Raw Materials for Manufacturing
Sigma-Aldrich Grades
Application Brand Grade or Trade Name Definition
Analytical
Fluka® Logo
TraceSELECT® Ultra Highest analytical grade for ultra-trace analysis. Designed for the ultra-trace analysis at ppb and even ppt levels. Typical metal trace impurities are below 0.1 µg/kg (<0.1 ppb), in some cases even as low as the ppq level.
Analytical
Fluka® Logo
TraceSELECT® High purity reagents for inorganic trace analysis applications and organic residue analysis. Developed for sample preparation and analysis in the ppm and ppb level. The blank values for metal traces are typically below 0.01 mg/kg (0.01 ppm).
Analytical
Fluka® Logo
TraceCERT® Certified reference material according to ISO 17025 and ISO Guide 34. TraceCERT reference materials are developed and produced in an accredited laboratory fulfilling both ISO 17025 (general requirements for the competence of testing and calibration laboratories) and ISO Guide 34 (general requirements for the competence of reference material producers).
Life Science
Sigma® Life Science Logo
BioUltra Finest quality products for life science, with the most stringent testing protocols and highest purity. Superior manufacturing methods and product specific testing protocols are designed to ensure quality to our highest standard. Tests may include, but are not limited to, purity by gel electrophoresis, trace metal testing and application testing.
Life Science
Sigma® Life Science Logo
BioXtra Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo. Includes application-specific formulations, such as tablets and prepared solutions.
Life Science
Sigma® Life Science Logo
BioReagent Base grade products, suitability tested for specific applications such as molecular biology or cell culture.
Life Science
Sigma® Life Science Logo
% purity Sigma brand products used for basic life science applications.
Chemistry
Aldrich® Logo
Anhydrous High-purity solvents with extremely low water content specifically manufactured for moisture sensitive syntheses.
Chemistry
Aldrich® Logo
% purity High quality chemistry products defined by their assay value as a percent purity.
All Sigma-Aldrich® Logo
Fluka® Logo
CHROMASOLV® Solvents tested for suitability in HPLC applications. Highest quality solvents used in LC-MS and LC-NMR are branded Fluka.
All Sigma-Aldrich® Logo puriss p.a., ACS reagent Exceeds standard ACS specifications with additional trace metals analysis.
All Sigma-Aldrich® Logo ACS reagent Meets or exceeds ACS specifications. For general analysis and procedures requiring stringent quality specifications.
All Sigma-Aldrich® Logo Corresponds to official monograph (e.g. Ph.Eur., USP) Produced/procured, packaged and analyzed under appropriate quality systems for pharmaceutical applications according to the corresponding monograph.
All Sigma-Aldrich® Logo Meets analytical specifications of USP, FCC, NF, EP (Ph. Eur.), BP, and/or JP. Chemicals that meet the latest testing specifications of the United States Pharmacopeia (USP), Food Chemicals Codex (FCC), National Formulary (NF), Pharmacopeia Europe (EP or Ph. Eur.), British Pharmacopeia (BP), and/or Japanese Pharmacopeia (JP). For R&D and testing purposes only.
All Sigma-Aldrich® Logo ReagentPlus® or Laboratory reagent ≥95% for general laboratory use.
All Sigma-Aldrich® Logo Technical Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%. Variation in color and physical form is possible.
©2010 Sigma-Aldrich Co. All rights reserved. SIGMA, SAFC, SIGMA-ALDRICH, ALDRICH, FLUKA, and SUPELCO are trademarks belonging to Sigma-Aldrich Co. and its affiliate Sigma-Aldrich Biotechnology, L.P. Sigma brand products are sold through Sigma-Aldrich, Inc. Sigma-Aldrich, Inc. warrants that its products conform to the ­information contained in this and other Sigma-Aldrich publications. Purchaser must determine the suitability of the product(s) for their particular use. Additional terms and conditions may apply. Please see reverse side of the invoice or packing slip. TraceSELECT, TraceCERT, CHROMASOLV, Hydranal and ReagentPlus are registered trademarks of Sigma-Aldrich Biotechnology L.P. and Sigma-Aldrich Co. BCR and ERM are registered trademarks of the European Commission.