phone number
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
United Kingdom > Your UK Technical Services  > Brands and Grades
Brands and Grades
Description of Sigma Product Grades
Table Comparing Sigma and Competitor Product Grades
Links to Product Selection Guides on our Website
We will often have large number of different products available for particular chemical. These can be different brands e.g. made by Sigma, Aldrich or FLUKA, or different grades, suitable or perhaps specifically tested for differerent applications.
Please see the table below indicating the different grades that may be available and their intended area of use, along with the suggested equivalent VWR or Fisher grade.
Commonly Known as Sigma-Aldrich Grade VWR Grade Fisher Grade
Technical Techn Technical Technical
General Purpose Reagent Purum > 95.0% GPR SLR/Certified
  Puriss > 98.5%    
Analytical Reagent Purum p.a. Analar AR
  Puriss p.a.    
  Puriss p.a. plus    
Chemistry Applications ACS    
  SigmaUltra    
  ReagentPlus    
  Anhydrous    
Life Science Applications BioChemika* Molecular Biology Biotechnology
  * for Molecular Biology (MB)    
  * for Microbiology    
  * for Microscopy    
  * for Electrophoresis    
  * for Fluorescence    
  Cell Culture Grade*    
  * tested for compatibility with growth of cells in culture    
  Biotech*    
  * for Peptide synthesis    
  * for oligonucleotide synthesis    
  * for Protein sequencing    
  * for Electrophoresis    
  Biochemika Ultra    
  Biochemika Ultra for MB*    
  * tested for DNAse, RNAse,    
  Protease and Phosphatase    
Pharmaceutical Applications BP (British Pharmacopoeia)    
  USP (US Pharmacopoeia)    
  Ph.Eur (European Pharmacopoeia)    
  * for clinical research    
Chromatography Reagents CHROMOSOLV HiPersolv HPLC
  * for HPLC   GC
  * for GC    
Spectrometry Reagents SPECTRANAL Spectrosol Spectranalysed
  AAS Spectroscopy    
  * for UV    
  * for IR    
Traces Analysis Reagents TraceSelect (below 0.5 ppb) Aristar Primar
  TraceSelect Ultra (below 1 ppt)    
Volumetric Solutions Fixanal Convol Nomafona
Residue Analysis Reagents PESTANAL For Residue Analysis Distol
  OEKANAL    
Karl Fisher Reagents HYDRANAL    
Specials Chiraselect: Reagents for derivatisation of Chiral Molecules    
  Selectophore: Special ion-selective Molecules & aids for Electrode Production    
  Certified Reference Materials: For use as Primary or Secondary Reference Standards    
Links to Product Selection Guides Shown on Our Website
Please find below links to other parts of the Sigma-Aldrich website which give advice on choosing the grade of product appropriate for different applications.
Solvent Application Portal: This is a set of tables recommending the grade of solvent appropriate for use in analytical, chemistry, environmental and biochemical applications
Descriptions of the grades relevant to solvents, available from Sigma-aldrich are shown in this link.
Adsorbent selection guide
Agarose selection guide
Albumin selection guide
Antibiotic selection guide
Buffer reference centre
Cell lysis reference and selection guide
Detergent selection guide
GC column selection guide
His Select reference and selection guide
Life science product guides
Protease inhibitor cocktail selection guide
Syringe selection guide
Vial closure and septa selection guide