| Commonly Known as |
Sigma-Aldrich Grade |
VWR Grade |
Fisher Grade |
| Technical |
Techn |
Technical |
Technical |
| |
|
|
|
| General Purpose Reagent |
Purum > 95.0% |
GPR |
SLR/Certified |
| |
Puriss > 98.5% |
|
|
| |
|
|
|
| Analytical Reagent |
Purum p.a. |
Analar |
AR |
| |
Puriss p.a. |
|
|
| |
Puriss p.a. plus |
|
|
| |
|
|
|
| Chemistry Applications |
ACS |
|
|
| |
SigmaUltra |
|
|
| |
ReagentPlus |
|
|
| |
Anhydrous |
|
|
| |
|
|
|
| Life Science Applications |
BioChemika* |
Molecular Biology |
Biotechnology |
| |
* for Molecular Biology (MB) |
|
|
| |
* for Microbiology |
|
|
| |
* for Microscopy |
|
|
| |
* for Electrophoresis |
|
|
| |
* for Fluorescence |
|
|
| |
Cell Culture Grade* |
|
|
| |
* tested for compatibility with growth of cells in culture |
|
|
| |
Biotech* |
|
|
| |
* for Peptide synthesis |
|
|
| |
* for oligonucleotide synthesis |
|
|
| |
* for Protein sequencing |
|
|
| |
* for Electrophoresis |
|
|
| |
Biochemika Ultra |
|
|
| |
Biochemika Ultra for MB* |
|
|
| |
* tested for DNAse, RNAse, |
|
|
| |
Protease and Phosphatase |
|
|
| |
|
|
|
| Pharmaceutical Applications |
BP (British Pharmacopoeia) |
|
|
| |
USP (US Pharmacopoeia) |
|
|
| |
Ph.Eur (European Pharmacopoeia) |
|
|
| |
* for clinical research |
|
|
| |
|
|
|
| Chromatography Reagents |
CHROMOSOLV |
HiPersolv |
HPLC |
| |
* for HPLC |
|
GC |
| |
* for GC |
|
|
| |
|
|
|
| Spectrometry Reagents |
SPECTRANAL |
Spectrosol |
Spectranalysed |
| |
AAS Spectroscopy |
|
|
| |
* for UV |
|
|
| |
* for IR |
|
|
| |
|
|
|
| Traces Analysis Reagents |
TraceSelect (below 0.5 ppb) |
Aristar |
Primar |
| |
TraceSelect Ultra (below 1 ppt) |
|
|
| Volumetric Solutions |
Fixanal |
Convol |
Nomafona |
| |
|
|
|
| Residue Analysis Reagents |
PESTANAL |
For Residue Analysis |
Distol |
| |
OEKANAL |
|
|
| |
|
|
|
| Karl Fisher Reagents |
HYDRANAL |
|
|
| |
|
|
|
| Specials |
Chiraselect: Reagents for derivatisation of Chiral Molecules |
|
|
| |
Selectophore: Special ion-selective Molecules & aids for Electrode Production |
|
|
| |
Certified Reference Materials: For use as Primary or Secondary Reference Standards |