Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
High-performance HPLC Fittings and Interconnects
Demonstrates the proper use of our exclusive fittings designed for use with ultra-high pressure and narrow-bore HPLC columns. These finger-tight fittings are rated to 15,000 psi, and are specially designed to eliminate dead volume that contributes to peak broadening and decreased resolution.