Spectroquant® Prove Cleaning
Spectroquant® Prove is built to last and resistant to most chemicals used in drinking water analysis. In the event of spills, the cuvet receptacle can be removed for cleaning. There is also a drainage port in the instrument itself so that any remaining spills can be rinsed out with distilled water or methanol. The surface of the instrument is also safe to be used with common cleaners.