Analytical
Starch Testing Procedure using STA20 kit
The Starch Assay Kit is used for the quantitative, enzymatic determination of starch in food. The intensity of the pink color measured at 540 nm is proportional to the original glucose concentration.