|
||||
|
|
One of the unique features of the VersaFlash system is how quick and easy it is to change out cartridges between samples. Once you’ve processed the first sample, simply turn the handle on the VersaFlash station counterclockwise to the “idle” position and pull it out to release the spent cartridge. Then simply replace the spent cartridge with a new cartridge, push the handle in, and turn the handle clockwise to the “operate” position to seal the cartridge into the VersaFlash station. When working with the same size cartridges, cartridge change-out is fast and simple.
Troca Rápida do Cartucho do VersaPak |
![]() Request the VersaFlash CD which includes videos and more.
Order VersaFlash products through our online product catalog.
|
||