The GenElute Gel Extraction kit is designed for the rapid purification of 50 bp to 10 kb linear DNA fragments and plasmids from standard or low-melting agarose gels. This kit can also be used to purify DNA from polyacrylamide gels. The isolated DNA is suitable for a variety of downstream applications, such as automated DNA sequencing, PCR, restriction digestion, cloning, and labeling.
- Recoveries up to 80%
- Each column can bind up to 10 µg of DNA
- Up to 3.5 g can be processed per column
- Compatible with both standard and low-melting agarose in TAE or TBE buffer