Labware
Aldrich Chemical Dryer
The Aldrich Chemical Dryer dries samples using heat, vacuum and desiccant. This device offers precise digital temperature control with an enclosed heating block for safe operation.