Labware
Grab Rack & Grab Rack Plus from Heathrow Scientific
Silicone grips help hold 15mm tubes in place when the Grab Rack is tipped upside down. Grab Rack Plus comes with an ejector that simultaneously ejects all the tubes in the rack.