|
||||
|
|
The Aldrich Glass Lab Kit with threaded joints features greaseless, vacuum tight, threaded standard taper joints. The unique threaded nut with O-ring connectors in this system automatically separate ground-glass joints when unscrewed and reduced the chance of breakage. This Glassware kit is compatible with Aldrich Schlenk-type glassware components. |
|
||