Labware
Aldrich VerSAFunnels
The Aldrich VerSAFunnel is a modular Buchner funnel for maximum versatility and economy. Each VerSAFunnel component is interchangeable and replaceable permitting the use of a wide range of funnel porosities and capacities.
Features & Benefits:
  • Five interchangeable frit porosities: 4 microns up to 175 microns
  • PTFE cup gasket seals edge of frit during use
  • Replaceable components
Materials