|
||||
|
|
The EZview™ Red Affinity Gels have a vivid red dye that clearly defines the pellet from the supernatant. This enhanced visibility results in less tedious sample processing and greatly reduces the risk of pellet loss which translates to higher reproducibility and more accurate quantitation of the protein of interest. EZview™ Red Affinity Gels are available to isolate antibodies, and recombinantly tagged proteins. Immuno affinity resins include antibodies to FLAG, HA, and c-Myc tags. Pulldown of complexes that form with GST, poly-His, Streptavidin and Protein A & G resins are also available. |
|
||