Setting Up and Running TruPAGE™ Precast Gels
TruPAGE™ precast gels offer high rigidity for tear resistance and improved protein resolution so researchers can achieve quality separation the first time around. TruPAGE gel cassettes are compatible with popular gel electrophoresis equipment and have a hassle-free design, which includes extra tall wells to prevent lane to lane overflow, requires no tape and no comb removal, and does not require tools to open. Additionally, TruPAGE gels have an extended 2 year shelf life and are available at an affordable price compared to competitor's polyacrylamide gels.
To purchase TruPAGE, click here.