Skip to Content
AL
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Designed multiple ligands. An emerging drug discovery paradigm.
Designed multiple ligands. An emerging drug discovery paradigm.
Journal of medicinal chemistry (2005-10-14)
Richard Morphy, Zoran Rankovic
PMID
16220969
Product Description
P0778
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Pricing
C8856
Sigma-Aldrich
Captopril, meets USP testing specifications
Pricing
C4042
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Pricing