Merck
  • [Determination of diphenyl and o-phenylphenol in agricultural products by GC/MS].
[Determination of diphenyl and o-phenylphenol in agricultural products by GC/MS].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2009-01-22)
Kunihiko Takahashi, Masakazu Horie, Yoshihumi Hirose
ABSTRACT
A simple determination method of diphenyl (DP) and o-phenylphenol (OPP) in agricultural products by GC/MS was examined. DP and OPP were extracted with ethyl acetate in the presence of anh. sodium sulfate. After addition of n-butanol, the extract solution was concentrated. Clean-up was achieved by shaking with graphitized bulk carbon (Supelclean ENVI-Carb). Addition of polyethylene glycol sharpened the OPP peak on GC/MS analysis. The recoveries from 9 kinds of agricultural products spiked at 0.01 and 0.5 microg/g each were mostly in the range of 70 to 120%, except for 50% recovery of OPP from barley spiked at 0.01 microg/g. The quantification limits (S/N > or =10) of DP and OPP were 0.0013 and 0.005 microg/g (0.0025 and 0.01 microg/g in barley and soybean), respectively.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Sigma-Aldrich
2-Phenylphenol, 99%
Supelco
2-Phenylphenol, PESTANAL®, analytical standard