Merck
  • Synthesis and evaluation of several new (2-chloroethyl) nitrosocarbamates as potential anticancer agents
Synthesis and evaluation of several new (2-chloroethyl) nitrosocarbamates as potential anticancer agents
Journal of Medicinal Chemistry (2000)
Reynolds RC, et al.