All Photos(1)

52462

Supelco

Florisil® Applicability Test acc. to DIN EN ISO 9377-2/H53, Kit

Synonym(s):
H53 Florisil® application test kit, DIN EN ISO 9377-2/Florisil® application test kit
NACRES:
NA.21

grade

analytical standard

Quality Level

agency

DIN

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

Related Categories

Components

Kit contains:
Stearyl stearate
Mineral oil Standard mixture Typ A and B (~10 mg/mL in heptane)
Extraction Medium Stock Solution

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service