Merck
Millipore® Filter Membranes
Millipore cut disk filter
We manufacture a broad range of filter membranes in various pore size ratings and diameters to meet your needs. Whether you are filtering solvents, mobile phases, or solutions for HPLC and other chromatography applications or collecting and preparing samples for analysis, Millipore® filters offer consistent, reliable filtration for analytical methods. With over 70 years of expertise, we provide high quality filters for accurate, consistent results.
Sort by Relevance
Showing 1-20 of 455
Change View
Showing 1-20 of 455
Compare
Product Number
Product Name
Product Description
Pricing
Membrane Filter Characteristics
  • Chemical Compatibility: The filter material must be compatible with the chemical nature of the substance being filtered to avoid structural failure. It is important to consider both the compatibility of liquid samples and dissolved solutes that can interact with the membrane.
  • Wettability: For liquid filtration, the membrane must be wettable with the fluid being filtered. Hydrophilic membranes are easily wet with water and are preferred for filtering aqueous solutions. Hydrophobic membranes are recommended for gas filtration, low surface tension solvents, and venting, and can be wet in organic solvents such as methanol, allowing filtration of both aqueous liquids and organic solvents.
  • Pore Size: Pore size provides an indication of the largest pore diameter and can be related to the membrane’s ability to filter out particles of a certain size. Bubble point and bacterial retention testing are two commonly used methods for measuring pore size.
  • Diameter: The diameter, size, and shape of the filter are selected based on the apparatus used for filtration or sample collection.
  • Flow Rate: Defined as the time required for the flow stream to pass through the filter, flow rate may be measured for air or liquid. Flow rate generally decreases with smaller pore size, but altering membrane material, thickness, porosity, and pore architecture can lead to differences in flow rate.
  • Analyte Binding: Analyte binding refers to the loss of analytes during filtration, resulting in a filtrate with a different molecular composition than expected. Membranes with limited functionality (e.g. PVDF, PTFE) show very low analyte binding, whereas membranes with higher functionality (e.g. nylon, MCE) show a high level of analyte binding)
  • Optical Properties: When visually analyzing retentates, membrane optical properties must be compatible with the imaging method, such that the membrane provides a consistent background over the entire sample surface and does not impart additional noise during testing.
  • Extractables: Extractables are contaminants present in the final filtrate that originate in the filter or device. Filter extractables occur as three different types: shedding of filter materials or particulate extractables, residual chemicals from the manufacturing process, and surface modification chemistries washing off the filter. The presence of extractables can also be related to the chemical compatibility of the membrane with the solution being filtered. Generally, if a membrane is not chemically compatible with the solution, a higher level of extractables is observed in the filtrate.
  • Retentiveness: Retentiveness is the ability of a membrane to retain the particle or molecule of interest. 
Prefiltration and Depth Filters
  • Prefiltration: Prefiltration utilizes large pore membrane filters to remove large particulates, such as dirt or sediment, from samples prior to filtration with a smaller pore membrane filter. Using prefiltration in sample preparation can prevent premature filter clogging or fouling.
  • Depth filters: Depth filters retain particles internally rather than on the filter surface. Due to their high particle retention capacity, depth filters are frequently used for prefiltration.
  • Binders: Commonly used in non-woven, fiber-based materials, binders provide shape and strength to the final product. While binders are routinely used in glass fiber filters, these additives reduce thermal stability and can result in sample contamination by extractables.
  • Net Filters: With large and uniform pores, the net-like structure of net filters is used to remove large particulates (e.g. cells, proteins, dirt) for solution clarification of particulate analysis.
How to Choose a Millipore® Filter by Membrane Type
Scanning electron microscope image of a glass fiber filter membrane
Glass Fiber
  • Typically used to filter large particles or viscous solutions
  • Used in prefiltration, air monitoring, particle collection, scintillation counting, and gravimetric analysis
View Products
Scanning electron microscope image of a mixed cellulose esters (MCE) membrane
Mixed Cellulose Esters (MCE)
  • Produced from biologically inert cellulose acetate and cellulose nitrate
  • Most referenced general purpose membrane
View Products
SEM image of a nylon membrane Alt text: Scanning electron microscope image of a nylon membrane
Nylon
  • Broad chemical compatibility
  • Low PFAS extractables
View Products
Scanning electron microscope image of a polycarbonate membrane
Polycarbonate (PCTE)
  • Well-defined pores and a smooth transparent surface ideal for use with optical or electron microscopy
View Products
Scanning electron microscope image of a polyethersulfone (PES) membrane
Polyethersulfone (PES)
  • Millipore Express® PLUS PES exhibits fast flow rates and low protein binding
  • Thermal stability, durability, and resistance to acidic and alkaline solutions
  • Low PFAS extractables
View Products
Scanning electron microscope image of a polypropylene membrane
Polypropylene
  • High solvent-compatibility and thermal stability
  • Low PFAS extractables
View Products
Scanning electron microscope image of a polytetrafluoroethylene (PTFE) filter
Polytetrafluoroethylene (PTFE)
  • High strength, low extractables, broadest chemical compatibility
  • Hydrophilic PTFE is typically used for filtering aqueous solutions
  • Hydrophobic PTFE is typically used for filtering organic solvents and gasses
View Products
Scanning electron microscope image of a polyvinylidene chloride (PVC) membrane filter
Polyvinylidene Chloride (PVC)
  • Preferentially used for gravimetric analysis to quantify silica, carbon black, or quartz air particulates
  • Commonly used in air monitoring methods
View Products
Scanning electron microscope image of a polyvinylidene fluoride (PVDF) membrane filter
Polyvinylidene Fluoride (PVDF)
  • Hydrophilic Durapore® PVDF exhibits the lowest protein binding
  • Hydrophobic Durapore® PVDF exhibits high protein binding
View Products
Scanning electron microscope image of a quartz filter
Quartz
  • No surface filter reaction with acidic gasses
  • Inert
  • Well-suited for measuring heavy metal concentrations and small particle quantities
View Products
Scanning electron microscope image of a silver membrane filter
Silver
  • Highly resistant to thermal stress and aggressive chemicals
  • Low background for sensitive X-ray diffraction analysis.
  • Specified in many standardized air monitoring methods
View Products
Scanning electron microscope image of a Strat-M® synthetic membrane filter
Strat-M® Membrane
  • Synthetic, non-animal based model for transdermal diffusion testing
  • Predictive of diffusion in human skin without lot-to-lot variability, safety, or storage limitations
View Products
How to Choose a Millipore® Filter by Volume
The diameter of the filter is selected based on sample volume and should fit the apparatus or filter holder used for filtration.
Sample Volume
Recommended Filter Diameter
1 – 10 mL
13 mm
10 – 100 mL
25 - 33 mm
100 mL – 1 L
33 mm – 50 mm
1 – 8 L
>50 mm
How to Choose a Millipore® Filter by Pore Size
Pore size provides an indication of largest pore diameter and can be related to the membrane’s ability to filter out particles of a certain size.
Pore size
Application
0.025 to 0.05 µm
Microdialysis; trace element and small particulate analysis
0.1 µm
Mycoplasma removal
0.20 to 0.22 µm
Sterilizing filtration and ultracleaning of aqueous solutions and organic solvents (e.g., HPLC)
0.45 µm
Clarification of aqueous solutions and organic solvents
0.8 µm
Coarse particulate removal and removal of bacteria
10 µm
Removal of visible particles
100 µm
Removal of sand, activated carbon, bead resins
Related Resources
Page 1 of 2