Merck
Millex® Syringe Filters
An assortment of Millex<sup>®</sup> syringe filters in various colors and sizes, displayed against a white background. These filters represent a range of small-volume filtration options for laboratory use. The filters come in different colors such as blue, green, yellow, purple, and red, indicating different filter types or pore sizes. Each filter has a circular shape with a conical funnel-like structure on top for attaching to a syringe. The central part of the filters is white with labeling that includes the brand name “Millex®”. Some filters have a transparent top section while others are opaque. There is also one funnel accessory included in the image which is used to assist with pouring liquids into the filter. These filters set the standard in small-volume filtration (1-200 mL), and their unsurpassed consistency in sample preparation has led to the development of many methods specifying their use.
Setting the standard in small-volume filtration (1-200 mL), the unsurpassed consistency of sterile and nonsterile Millex® syringe filters in sample preparation has led to the development of many methods specifying their use. Our controlled manufacturing process ensures reliable filter performance. Featuring reduced sample loss due to minimal hold-up volumes, Millex® syringe filters are ideal for use in a variety of applications including:  
  • HPLC Sample Preparation
  • Dissolution Testing
  • Hard-to-filter, Nonsterile Samples
  • Filtration of Antibiotics and Tissue Culture Additives
  • Medical Applications
  • Air and Gas Venting with Hydrophobic PTFE Membranes
Products
non-sterile (86)
sterile (58)
0.45 (58)
0.22 (39)
0.2 (31)
0.8 (5)
5.0 (4)
0.1 (3)
PTFE (46)
PVDF (42)
polyethersulfone (20)
mixed cellulose esters (MCE) (18)
Nylon (15)
glass fiber (3)
Sort by Relevance
Showing 1-20 of 144
Change View
Showing 1-20 of 144
Millex<SUP>™</SUP> PVDF syringe filter
SLHV033N
Millex PVDF syringe filter
pore size 0.45 μm, diam. 33 mm, non-sterile, hydrophilic
Millex<SUP>™</SUP> polyethersulfone syringe filter
SLGPR33RS
Millex polyethersulfone syringe filter
pore size 0.22 μm, diam. 33 mm, sterile, hydrophilic
Millex<SUP>™</SUP>-GP Filter Unit (Sterile)
SLGP033R
Millex-GP Filter Unit (Sterile)
pore size 0.22 μm, diam. 33 mm, polyethersulfone membrane, hydrophilic, medical device
Millex<SUP>™</SUP> PVDF syringe filter
SLGVR33RS
Millex PVDF syringe filter
pore size 0.22 μm, diam. 33 mm, sterile, hydrophilic
Millex<SUP>™</SUP>-GV Filter Unit (Sterile)
SLGV033R
Millex-GV Filter Unit (Sterile)
pore size 0.22 μm, diam. 33 mm, PVDF membrane, hydrophilic, medical device
Vented Millex<SUP>™</SUP>-GV Filter Unit (Sterile)
SLGVV255F
Vented Millex-GV Filter Unit (Sterile)
pore size 0.22 μm, diam. 25 mm, PVDF membrane, hydrophilic, medical device
Millex<SUP>™</SUP>-GP Filter Unit (Sterile)
SLGPM33RS
Millex-GP Filter Unit (Sterile)
pore size 0.22 μm, diam. 33 mm, polyethersulfone membrane, hydrophilic, medical device
Millex<SUP>™</SUP> PVDF syringe filter
SLGV004SL
Millex PVDF syringe filter
pore size 0.22 μm, diam. 4 mm, sterile, hydrophilic
Millex<SUP>™</SUP> polyethersulfone syringe filter
SLGPR33RB
Millex polyethersulfone syringe filter
pore size 0.22 μm, diam. 33 mm, sterile, hydrophilic
Millex<SUP>™</SUP> hydrophobic PTFE vent filter
SLFG050
Millex hydrophobic PTFE vent filter
pore size 0.2 μm, diam. 50 mm, non-sterile, 1/4-1/2 in. hose barb, sterilization compatible with autoclavable
Millex<SUP>™</SUP> PVDF syringe filter
SLHVR33RS
Millex PVDF syringe filter
pore size 0.45 μm, diam. 33 mm, sterile, hydrophilic
Millex<SUP>™</SUP>-GV Filter Unit (Sterile)
SLGVM33RS
Millex-GV Filter Unit (Sterile)
pore size 0.22 μm, diam. 33 mm, PVDF membrane, hydrophilic, medical device
Millex<SUP>™</SUP> hydrophilic PTFE syringe filter
SLCR033
Millex hydrophilic PTFE syringe filter
pore size 0.45 μm, diam. 33 mm, non-sterile
Millex<SUP>™</SUP> hydrophilic PTFE syringe filter
SLLG033
Millex hydrophilic PTFE syringe filter
pore size 0.20 μm, diam. 33 mm, non-sterile
Millex<SUP>™</SUP>-GV Filter Unit (Sterile)
SLGV013SL
Millex-GV Filter Unit (Sterile)
pore size 0.22 μm, diam. 13 mm, PVDF membrane, hydrophilic, medical device
Millex<SUP>™</SUP>-FG Filter Unit (Sterile)
SLFG025LS
Millex-FG Filter Unit (Sterile)
pore size 0.2 μm, diam. 25 mm, PTFE membrane, hydrophobic, medical device
Millex<SUP>™</SUP> Nylon syringe filter
SLHN033
Millex Nylon syringe filter
pore size 0.45 μm, diam. 33 mm, non-sterile, hydrophilic
Millex<SUP>™</SUP> PVDF syringe filter
SLGV033N
Millex PVDF syringe filter
pore size 0.22 μm, diam. 33 mm, non-sterile, hydrophilic
Millex<SUP>™</SUP>-GS Filter Unit (Sterile)
SLGS033
Millex-GS Filter Unit (Sterile)
pore size 0.22 μm, diam. 33 mm, mixed cellulose esters (MCE) membrane, hydrophilic, medical device
Millex<SUP>™</SUP> PVDF syringe filter
SLGVX13
Millex PVDF syringe filter
pore size 0.22 μm, diam. 13 mm, non-sterile, hydrophilic
Not finding the right product?  
Try our Product Selector Tool.
Sterile Millex® Syringe Filters
  • Applications include sterilizing cell culture reagents such as cell culture media, antibiotics, supplements, sera, viruses for infection, and other biological solutions
  • Fast flow and low binding for cell culture media preparation with Millipore Express® PLUS polyethersulfone (PES) membrane
  • Durapore® PVDF membrane is the lowest binding membrane for protein-rich solutions
  • Many are CE-marked and/or FDA registered for medical intended use
  • Minimal leaching of impurities into filtered solutions
  • 50 mm Millex® syringe filters have hydrophobic chemistry for gas filtration and in-line vacuum pump protection, and are autoclavable
Non-sterile Millex® Syringe Filters
  • Applications include filtration-based sample preparation for dissolution testing, HPLC, UHPLC, LC-MS, ion chromatography (IC), and general particulate removal
  • Hydrophilic PTFE Millex® filters are HPLC-certified for low extractables
  • Best recovery of protein samples with hydrophilic Durapore® PVDF Millex® syringe filters
  • Automation-compatible, non-sterile Millex® syringe filter units have a domed housing which enables smooth, reliable delivery in automated sample preparation workstations by eliminating shingling between filters on the transport rack.
  • Nonsterile HPF Millex® filters include a graduated glass fiber prefilter to remove larger particles and either a 0.2 µm or 0.45 µm membrane filter for fine filtration. This combination of prefilter and membrane provides significantly greater throughput than standard filters without prefiltration media, especially when filtering particle laden solutions.
  • 50 mm Millex® syringe filters have hydrophobic chemistry for gas filtration and in-line vacuum pump protection
How to Choose Millex Filters® by Membrane Type
Zoomed photograph of a 33 mm Millex<sup>®</sup> syringe filter with Durapore® PVDF membrane
Durapore® PVDF Membrane
  • Lowest binding
View Products
Zoomed photograph of a 33 mm Millex<sup>®</sup> syringe filter with Millipore Express<sup>®</sup> PLUS PES membrane
Millipore Express® PLUS Polyethersulfone (PES) Membrane
  • Fast flow rates and low protein binding for cell culture media preparation.
View Products
Zoomed photograph of two 33 mm Millex<sup>®</sup> syringe filters with hydrophilic PTFE membrane
Hydrophilic PTFE Membrane
  • Lowest HPLC-UV extractables with broadest chemical compatibility
View Products
Zoomed photograph of two 33 mm Millex<sup>®</sup> syringe filters with hydrophobic PTFE membrane
Hydrophobic PTFE Membrane
  • Excellent solvent resistance; vent applications
View Products
Zoomed photograph of a 33 mm Millex<sup>®</sup> syringe filter with nylon membrane
Nylon Membrane
  • Broad chemical compatibility
View Products
Zoomed photograph of a 33 mm Millex<sup>®</sup> syringe filter with mixed cellulose esters (MCE) membrane
MCE (mixed cellulose esters) Membrane
  • Most referenced general purpose membrane
View Products
Zoomed photograph of two 33 mm Millex<sup>®</sup> syringe filters with glass fiber filter
Glass Fiber Filter
  • Clarification of solutions containing high particulate levels
View Products
Cartoon illustration of a Millex<sup>®</sup> syringe filter, with a double-headed arrow underneath indicating the "diameter" parameter
How to choose Millex® filters by sample volume
Sample VolumeMillex® syringe filter diameter
<1 mL
4 mm
1 – 10 mL
13 mm
10 – 100 mL
25 – 33 mm
4 – 8 L
50 mm
Related Resources
Related Webinars
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Filtration in Dissolution Testing: Improving Throughput and Reducing Variability
In vitro dissolution testing is used to characterize drug compounds throughout their development. In early drug development it is used to support the choice of a particular formulation. During drug production it is a critical component of the quality control process and is used to assess the changes in manufacturing processes or formulation. In order for dissolution results to be meaningful at each stage, the test and the process need to be reliable, consistent, predictive and accurate.