Recombinant/Fusion Tag Protein Purification
Recombinant protein purification by tag-specific affinity chromatography is a proven technology that results in highly specific recognition and purification of recombinant proteins. The expression, purification, and detection of recombinant proteins can be a lengthy and time-consuming endeavor. Depending on the protein of interest and expression system, protein tags may be added to improve the outcome of common steps including protein solubility during expression, purification by affinity chromatography, or immunodetection following isolation and purification. We offer affinity columns and resins for multiple fusion tags in numerous formats, including magnetic beads, packed columns, and agarose resin slurries, ensuring we have products for all various scales and applications.
Products
Anti-FLAG® M2 Magnetic Beads
affinity isolated antibody
3xFLAG™ Peptide
≥90% (HPLC/MS), lyophilized powder
d-Desthiobiotin
≥98% (TLC)
FLAG® Peptide
≥85% (HPLC), lyophilized powder
HIS-Select® Nickel Affinity Gel
(1:1 suspension in a 20% ethanol solution)
Anti-HA Magnetic Beads
Magnetic agarose, suspension
Glutathione−Agarose
lyophilized powder
Glutathione High Capacity Magnetic Agarose Beads
(1:1 suspension in a 30% ethanol solution)
HIS-Select® Nickel Magnetic Agarose Beads
HIS-Select® HF Nickel Affinity Gel
Anti-C-Myc Magnetic Beads
Magnetic agarose, suspension
HIS-Select® Spin Columns
FLAG® M Purification Kit
For Mammalian expression systems.
PureProteome Nickel Magnetic Bead System
PureProteome Nickel Magnetic Beads provide researchers with a powerful bead system to help purify polyhistidine-tagged recombinant proteins
HIS-Select® Cobalt Affinity Gel
(1:1 suspension in a 30% ethanol solution)
HIS-Select® Elution Buffer
Anti-MBP Magnetic Beads
Magnetic agarose, suspension
EZview™ Red Glutathione Affinity Gel
Glutathione−Agarose
set of 3 pre-packed columns (2.5 ml each), (1:1 suspension in a 0.5 M NaCl + 20% ethanol solution)
HIS-Select® Filter Plate
Cytiva™ Amersham Chromatography Columns and Media
A wide range of Cytiva™ Amersham chromatography media is available to purify proteins using either manual or automated methods. Prepacked formats include gravity columns, spin columns, 96-well plates, and prepacked columns for use with automated chromatography systems.
- His-tagged proteins: Fast and convenient histidine-tagged recombinant protein purification using immobilized metal ion affinity chromatography (IMAC). Ni Sepharose media are precharged with nickel ions (Ni2+); TALON® Superflow™ medium is precharged with cobalt ions (Co2+).
- GST-tagged proteins: Glutathione Sepharose chromatography media are available in lab packs, prepacked GSTrap and GSPrep columns, and 96-well plates. GST Purification Modules are offered for purification of bacterial lysates. GST SpinTrap modules includes prepacked Glutathione Sepharose 4B SpinTrap columns as well as the buffers needed for purification. RediPack GST Purification Modules include gravity flow columns and buffer.
- Strep-tag™ II-tagged proteins: Strep-tag II binds specifically to StrepTactin™ Sepharose High Performance resin, which has StrepTactin ligand immobilized on a Sepharose base matrix to yield pure target protein.
- MBP-tagged proteins: Dextrin Sepharose High Performance chromatography medium purifies recombinant proteins tagged with maltose binding protein (MBP).
Agarose Beads and Affinity Gels
Agarose beads and affinity gels enable fusion-tagged protein purification using ligands cross-linked to beaded agarose. Good for small to large scale purification under native, denaturing, or mildly reducing conditions, these resins are suitable for gravity flow column purification, low-speed centrifugation in batch processing, and low-pressure chromatography procedures as the beads and gels are designed for lower pressures. EZview™ affinity gels incorporate a red dye that allows the user to easily differentiate pellet from supernatant, resulting in less protein loss during batch purification. Immunoprecipitation kits include affinity gels, mini-spin columns, and lysis reagent and are specially designed to allow maximal recovery of proteins from immunoprecipitates.
Superflow Agarose and High-flow Affinity Gels
Superflow agarose and High-flow affinity gels use highly crosslinked agarose beads that can withstand higher pressures compared to standard agarose bead resins. This makes these resins suitable for fast protein liquid chromatography (FPLC) purification methods. Superflow agarose and high-flow affinity gels are recommended for medium to large scale purification.
Magnetic Beads
Magnetic beads are used to purify recombinant fusion tagged protein using a magnetic rack or platform to separate beads from wash and elution fractions. This procedure allows for rapid processing under native or denaturing conditions within seconds, and is suitable for low throughput using single microfuge tubes or higher throughput using 96-well plates and automation.
BugBuster® Purification Kits
BugBuster® purification kits combine affinity resin, wash buffers, elution buffers, and extraction reagent for convenient preparation of soluble cell extracts and affinity purification of tagged fusion proteins from E. coli.
Related Resources
- Brochure: Refine Protein Preparation - Tools for Better Protein Analysis
Today, researchers are challenged to create high quality samples for meaningful protein analysis, often using cumbersome traditional sample preparation methods. With over 50 years of experience in developing protein sample preparation technologies, MilliporeSigma is constantly innovating new tools to offer you rapid and efficient solutions that can be smoothly integrated into your workflow.
- Catalog: Recombinant Proteins – Purification and Detection Products
Why spend your time on arduous sample preparation protocols when you can focus your efforts on exciting experiments? With the right pure protein, in the buffer you need, at the concentration you want, your next discovery is only a step away. From protein isolation to purification, you can count on us to support your research with maximum yields of intact, functional proteins..