Skip to Content
Merck
Formaldehyde Solution 36%

Formaldehyde Solution

Formaldehyde Solution 36%
Synonym(s):
Formaldehyde Solution, Formalin, Formaldehyde
Empirical Formula (Hill Notation):
CH2O
Molecular Weight:
30.03

Products

Brand
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing