Skip to Content
Merck
Water with 0.1% (v/v) Formic acid hypergrade for LC-MS LiChrosolv®

Water with 0.1% (v/v) Formic acid

Water with 0.1% (v/v) Formic acid hypergrade for LC-MS LiChrosolv®
Synonym(s):
water solution; mobile phase

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-2 of 2